Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 73199818-73200051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG1
Entrez Gene ID
9382
Gene Name
component of oligomeric golgi complex 1
Gene Aliases
CDG2G, LDLB
UniGene
Hs.103555
Gene Chromosome Location
Chr.17: 73193034-73208507 on Build GRCh38
Transcripts
NM_018714.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6556627 c.984C>T p.I328I Chr.17: 73199935-73199935 on Build GRCh38
6241349 c.1006C>A p.R336R Chr.17: 73199957-73199957 on Build GRCh38
6367589 c.994C>T p.Q332* Chr.17: 73199945-73199945 on Build GRCh38
5743863 c.1007G>A p.R336Q Chr.17: 73199958-73199958 on Build GRCh38
4167805 c.1037A>G p.Y346C Chr.17: 73199988-73199988 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140736202 Chr.17: 73199876-73199876 on Build GRCh38
rs779797550 Chr.17: 73200005-73200005 on Build GRCh38
rs766581270 Chr.17: 73199869-73199869 on Build GRCh38
rs774720405 Chr.17: 73199962-73199962 on Build GRCh38
rs753327660 Chr.17: 73199933-73199933 on Build GRCh38
rs959200300 Chr.17: 73199872-73199872 on Build GRCh38
rs773341615 Chr.17: 73199957-73199957 on Build GRCh38
rs970113883 Chr.17: 73199952-73199952 on Build GRCh38
rs763005617 Chr.17: 73199958-73199958 on Build GRCh38
rs764574310 Chr.17: 73199978-73199978 on Build GRCh38
rs761810775 Chr.17: 73199857-73199857 on Build GRCh38
rs756198029 Chr.17: 73199917-73199918 on Build GRCh38
rs779924491 Chr.17: 73199939-73199939 on Build GRCh38
rs777314683 Chr.17: 73199919-73199919 on Build GRCh38
rs775416328 Chr.17: 73199846-73199846 on Build GRCh38
rs911276293 Chr.17: 73200007-73200007 on Build GRCh38
rs150109830 Chr.17: 73199891-73199891 on Build GRCh38
rs781005411 Chr.17: 73200030-73200030 on Build GRCh38
rs761212185 Chr.17: 73199975-73199975 on Build GRCh38
rs562332952 Chr.17: 73199953-73199953 on Build GRCh38
rs752833886 Chr.17: 73199849-73199849 on Build GRCh38
rs201281586 Chr.17: 73200024-73200024 on Build GRCh38
rs755527594 Chr.17: 73199917-73199917 on Build GRCh38
rs763206860 Chr.17: 73199868-73199868 on Build GRCh38
rs375438969 Chr.17: 73199865-73199865 on Build GRCh38
rs768071925 Chr.17: 73199879-73199879 on Build GRCh38
rs751959598 Chr.17: 73199871-73199871 on Build GRCh38
rs769941790 Chr.17: 73199862-73199862 on Build GRCh38
rs768018542 Chr.17: 73199967-73199967 on Build GRCh38
rs142802630 Chr.17: 73200002-73200002 on Build GRCh38
rs745601566 Chr.17: 73199937-73199937 on Build GRCh38
rs771219802 Chr.17: 73199960-73199960 on Build GRCh38
rs771855863 Chr.17: 73199938-73199938 on Build GRCh38
rs759941239 Chr.17: 73199964-73199964 on Build GRCh38
rs778591192 Chr.17: 73199936-73199936 on Build GRCh38
rs4375725 Chr.17: 73200020-73200020 on Build GRCh38
rs746631479 Chr.17: 73200006-73200006 on Build GRCh38
rs776043683 Chr.17: 73199969-73199969 on Build GRCh38
rs756727694 Chr.17: 73199988-73199988 on Build GRCh38
rs377061582 Chr.17: 73200016-73200016 on Build GRCh38
rs117344829 Chr.17: 73200000-73200000 on Build GRCh38
rs369505856 Chr.17: 73199918-73199918 on Build GRCh38
rs199576608 Chr.17: 73199986-73199986 on Build GRCh38
rs746945599 Chr.17: 73199941-73199941 on Build GRCh38
rs756930348 Chr.17: 73199935-73199935 on Build GRCh38
rs764799546 Chr.17: 73199991-73199991 on Build GRCh38

View Full Product Details