Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 14908974-14909213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCLRE1C
Entrez Gene ID
64421
Gene Name
DNA cross-link repair 1C
Gene Aliases
A-SCID, DCLREC1C, RS-SCID, SCIDA, SNM1C
UniGene
Hs.655932
Gene Chromosome Location
Chr.10: 14897853-14954432 on Build GRCh38
Transcripts
NM_001033855.2 NM_001033857.2 NM_001033858.2 NM_001289076.1 NM_001289077.1 NM_001289078.1 NM_001289079.1 NM_022487.3 NR_110297.1 XM_006717491.3 XM_011519616.1 XM_011519617.1 XM_011519619.1 XM_017016556.1 XM_017016557.1 XM_017016558.1 XM_017016559.1 XM_017016560.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
916629 c.1126G>T p.V376L Chr.10: 14909016-14909016 on Build GRCh38
4333117 c.1458A>G p.V486V Chr.10: 14909029-14909029 on Build GRCh38
300467 c.1385_1387delAAG p.E462delE Chr.10: 14909100-14909102 on Build GRCh38
4595707 c.1115C>G p.P372R Chr.10: 14909027-14909027 on Build GRCh38
325128 c.1045G>A p.G349R Chr.10: 14909097-14909097 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs921819288 Chr.10: 14909184-14909184 on Build GRCh38
rs369306346 Chr.10: 14909173-14909173 on Build GRCh38
rs80148779 Chr.10: 14909147-14909147 on Build GRCh38
rs1056418621 Chr.10: 14909066-14909066 on Build GRCh38
rs746063292 Chr.10: 14909125-14909125 on Build GRCh38
rs933736163 Chr.10: 14909138-14909138 on Build GRCh38
rs771454743 Chr.10: 14909116-14909116 on Build GRCh38
rs144654282 Chr.10: 14909119-14909119 on Build GRCh38
rs772499261 Chr.10: 14909023-14909023 on Build GRCh38
rs757055066 Chr.10: 14909070-14909070 on Build GRCh38
rs780097801 Chr.10: 14909073-14909075 on Build GRCh38
rs755790137 Chr.10: 14909170-14909170 on Build GRCh38
rs747435874 Chr.10: 14909032-14909032 on Build GRCh38
rs116202790 Chr.10: 14909028-14909028 on Build GRCh38
rs483352702 Chr.10: 14909145-14909145 on Build GRCh38
rs35599500 Chr.10: 14909157-14909158 on Build GRCh38
rs376186052 Chr.10: 14909153-14909153 on Build GRCh38
rs750613031 Chr.10: 14909008-14909009 on Build GRCh38
rs766009897 Chr.10: 14909174-14909174 on Build GRCh38
rs963509408 Chr.10: 14909159-14909159 on Build GRCh38
rs764005111 Chr.10: 14909052-14909052 on Build GRCh38
rs753674918 Chr.10: 14909172-14909172 on Build GRCh38
rs373709012 Chr.10: 14909100-14909102 on Build GRCh38
rs368636876 Chr.10: 14909076-14909076 on Build GRCh38
rs755774055 Chr.10: 14909100-14909100 on Build GRCh38
rs775633789 Chr.10: 14909146-14909146 on Build GRCh38
rs746552030 Chr.10: 14909106-14909106 on Build GRCh38
rs751338558 Chr.10: 14909061-14909061 on Build GRCh38
rs765379729 Chr.10: 14909048-14909048 on Build GRCh38
rs756239667 Chr.10: 14909009-14909011 on Build GRCh38
rs778156435 Chr.10: 14909108-14909108 on Build GRCh38
rs894826033 Chr.10: 14909068-14909068 on Build GRCh38
rs747311091 Chr.10: 14909111-14909111 on Build GRCh38
rs761553201 Chr.10: 14909156-14909156 on Build GRCh38
rs374914377 Chr.10: 14909165-14909165 on Build GRCh38
rs776999438 Chr.10: 14909037-14909037 on Build GRCh38
rs759650664 Chr.10: 14909041-14909041 on Build GRCh38
rs954715514 Chr.10: 14909191-14909191 on Build GRCh38
rs1013182702 Chr.10: 14909072-14909072 on Build GRCh38
rs750104684 Chr.10: 14909079-14909079 on Build GRCh38
rs904814123 Chr.10: 14909022-14909022 on Build GRCh38
rs748820739 Chr.10: 14909103-14909103 on Build GRCh38
rs567152555 Chr.10: 14909162-14909162 on Build GRCh38
rs1046631 Chr.10: 14909114-14909114 on Build GRCh38
rs774273800 Chr.10: 14909154-14909154 on Build GRCh38
rs753332017 Chr.10: 14909181-14909181 on Build GRCh38
rs786200884 Chr.10: 14909135-14909141 on Build GRCh38
rs115250914 Chr.10: 14909102-14909102 on Build GRCh38
rs950457999 Chr.10: 14909110-14909110 on Build GRCh38
rs769948633 Chr.10: 14909143-14909143 on Build GRCh38

View Full Product Details