Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 31091582-31091843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX11
Entrez Gene ID
1663
Gene Name
DEAD/H-box helicase 11
Gene Aliases
CHL1, CHLR1, KRG2, WABS
UniGene
Hs.443960
Gene Chromosome Location
Chr.12: 31073574-31104799 on Build GRCh38
Transcripts
NM_001257144.1 NM_001257145.1 NM_004399.2 NM_030653.3 NM_152438.1 XM_005253331.3 XM_005253333.3 XM_006719050.3 XM_011520592.2 XM_011520595.2 XM_011520597.2 XM_017018918.1 XM_017018919.1 XM_017018920.1 XM_017018921.1 XM_017018922.1 XM_017018923.1 XM_017018924.1 XM_017018925.1 XM_017018926.1 XM_017018927.1 XM_017018928.1 XM_017018929.1 XM_017018930.1 XM_017018931.1 XM_017018932.1 XM_017018933.1 XM_017018934.1 XM_017018935.1 XM_017018936.1 XM_017018937.1 XM_017018938.1 XM_017018939.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6136812 c.1210G>T p.G404C Chr.12: 31091839-31091839 on Build GRCh38
938688 c.1142C>T p.A381V Chr.12: 31091771-31091771 on Build GRCh38
2205299 c.1208C>T p.T403M Chr.12: 31091837-31091837 on Build GRCh38
4041378 c.1198G>A p.D400N Chr.12: 31091827-31091827 on Build GRCh38
6641530 c.1151G>A p.R384Q Chr.12: 31091780-31091780 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61917221 Chr.12: 31091685-31091685 on Build GRCh38
rs4031292 Chr.12: 31091667-31091667 on Build GRCh38
rs150466604 Chr.12: 31091814-31091814 on Build GRCh38
rs754864339 Chr.12: 31091759-31091759 on Build GRCh38
rs192257386 Chr.12: 31091767-31091767 on Build GRCh38
rs778677044 Chr.12: 31091760-31091760 on Build GRCh38
rs78383308 Chr.12: 31091772-31091772 on Build GRCh38
rs752441260 Chr.12: 31091813-31091813 on Build GRCh38
rs771639157 Chr.12: 31091709-31091709 on Build GRCh38
rs776195047 Chr.12: 31091733-31091733 on Build GRCh38
rs751043573 Chr.12: 31091826-31091826 on Build GRCh38
rs368266910 Chr.12: 31091762-31091762 on Build GRCh38
rs754879885 Chr.12: 31091686-31091686 on Build GRCh38
rs776890809 Chr.12: 31091725-31091725 on Build GRCh38
rs765068084 Chr.12: 31091809-31091809 on Build GRCh38
rs757234056 Chr.12: 31091732-31091732 on Build GRCh38
rs573826946 Chr.12: 31091806-31091806 on Build GRCh38
rs74439745 Chr.12: 31091769-31091769 on Build GRCh38
rs777235655 Chr.12: 31091710-31091710 on Build GRCh38
rs745493626 Chr.12: 31091770-31091770 on Build GRCh38
rs751663827 Chr.12: 31091753-31091753 on Build GRCh38
rs752674196 Chr.12: 31091761-31091761 on Build GRCh38
rs899309959 Chr.12: 31091699-31091699 on Build GRCh38
rs563109212 Chr.12: 31091805-31091805 on Build GRCh38
rs763952460 Chr.12: 31091823-31091823 on Build GRCh38
rs761977000 Chr.12: 31091754-31091754 on Build GRCh38
rs9788173 Chr.12: 31091658-31091658 on Build GRCh38
rs759877549 Chr.12: 31091726-31091726 on Build GRCh38
rs938061376 Chr.12: 31091615-31091615 on Build GRCh38
rs764305418 Chr.12: 31091748-31091748 on Build GRCh38
rs201612562 Chr.12: 31091731-31091731 on Build GRCh38
rs746714944 Chr.12: 31091717-31091717 on Build GRCh38
rs150966927 Chr.12: 31091771-31091771 on Build GRCh38
rs753591664 Chr.12: 31091756-31091756 on Build GRCh38
rs397842879 Chr.12: 31091755-31091755 on Build GRCh38
rs770950577 Chr.12: 31091809-31091811 on Build GRCh38
rs770569119 Chr.12: 31091721-31091721 on Build GRCh38
rs372573378 Chr.12: 31091779-31091779 on Build GRCh38
rs372111156 Chr.12: 31091700-31091700 on Build GRCh38
rs779758501 Chr.12: 31091670-31091670 on Build GRCh38
rs35588475 Chr.12: 31091782-31091782 on Build GRCh38
rs149773582 Chr.12: 31091780-31091780 on Build GRCh38
rs763547126 Chr.12: 31091735-31091735 on Build GRCh38
rs767483820 Chr.12: 31091803-31091803 on Build GRCh38
rs778767280 Chr.12: 31091692-31091692 on Build GRCh38
rs993681121 Chr.12: 31091797-31091797 on Build GRCh38
rs759797718 Chr.12: 31091650-31091650 on Build GRCh38
rs761898940 Chr.12: 31091784-31091784 on Build GRCh38

View Full Product Details