Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42980393-42980666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTGES3L
Entrez Gene ID
100885848
Gene Name
prostaglandin E synthase 3 like
Gene Aliases
-
UniGene
Hs.317403
Gene Chromosome Location
Chr.17: 42968088-42980528 on Build GRCh38
Transcripts
NM_001142653.1 NM_001142654.1 NM_001261430.1 NM_001136042.2 NM_001321381.1 NM_173079.3 XM_005257078.3 XM_005257080.3

Gene Symbol
PTGES3L-AARSD1
Entrez Gene ID
100885850
Gene Name
PTGES3L-AARSD1 readthrough
Gene Aliases
-
UniGene
Hs.317403
Gene Chromosome Location
Chr.17: 42950526-42980528 on Build GRCh38
Transcripts
NM_001142653.1 NM_001142654.1 NM_001261430.1 NM_001136042.2 NM_001321381.1 NM_173079.3 XM_005257078.3 XM_005257080.3

Gene Symbol
RUNDC1
Entrez Gene ID
146923
Gene Name
RUN domain containing 1
Gene Aliases
-
UniGene
Hs.632255
Gene Chromosome Location
Chr.17: 42980510-42993692 on Build GRCh38
Transcripts
NM_001142653.1 NM_001142654.1 NM_001261430.1 NM_001136042.2 NM_001321381.1 NM_173079.3 XM_005257078.3 XM_005257080.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6770598 c.34A>T p.T12S Chr.17: 42980610-42980610 on Build GRCh38
390785 c.35C>A p.T12K Chr.17: 42980611-42980611 on Build GRCh38
5031629 c.72A>C p.E24D Chr.17: 42980648-42980648 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752347491 Chr.17: 42980625-42980625 on Build GRCh38
rs894095641 Chr.17: 42980574-42980574 on Build GRCh38
rs753362536 Chr.17: 42980608-42980608 on Build GRCh38
rs888347296 Chr.17: 42980417-42980417 on Build GRCh38
rs754932911 Chr.17: 42980506-42980506 on Build GRCh38
rs746367589 Chr.17: 42980590-42980590 on Build GRCh38
rs774649733 Chr.17: 42980579-42980590 on Build GRCh38
rs1033457519 Chr.17: 42980474-42980474 on Build GRCh38
rs692074 Chr.17: 42980561-42980561 on Build GRCh38
rs764334670 Chr.17: 42980599-42980599 on Build GRCh38
rs763219665 Chr.17: 42980526-42980526 on Build GRCh38
rs971007167 Chr.17: 42980428-42980428 on Build GRCh38
rs770339301 Chr.17: 42980592-42980592 on Build GRCh38
rs964932296 Chr.17: 42980431-42980431 on Build GRCh38
rs973194695 Chr.17: 42980444-42980444 on Build GRCh38
rs750401321 Chr.17: 42980555-42980555 on Build GRCh38
rs900249593 Chr.17: 42980615-42980615 on Build GRCh38
rs949513235 Chr.17: 42980525-42980525 on Build GRCh38
rs754479522 Chr.17: 42980613-42980613 on Build GRCh38
rs867551579 Chr.17: 42980499-42980499 on Build GRCh38
rs1042094571 Chr.17: 42980600-42980600 on Build GRCh38
rs374885370 Chr.17: 42980627-42980627 on Build GRCh38
rs764271621 Chr.17: 42980545-42980545 on Build GRCh38
rs750311395 Chr.17: 42980604-42980604 on Build GRCh38
rs945576436 Chr.17: 42980584-42980584 on Build GRCh38
rs755621160 Chr.17: 42980558-42980558 on Build GRCh38
rs17853899 Chr.17: 42980586-42980586 on Build GRCh38
rs748808612 Chr.17: 42980564-42980564 on Build GRCh38
rs778582103 Chr.17: 42980618-42980618 on Build GRCh38
rs1015393984 Chr.17: 42980427-42980427 on Build GRCh38
rs562961994 Chr.17: 42980583-42980583 on Build GRCh38
rs1049018389 Chr.17: 42980611-42980611 on Build GRCh38
rs541258538 Chr.17: 42980492-42980492 on Build GRCh38
rs978781953 Chr.17: 42980530-42980530 on Build GRCh38
rs781518919 Chr.17: 42980643-42980643 on Build GRCh38
rs1004540313 Chr.17: 42980423-42980423 on Build GRCh38
rs199576891 Chr.17: 42980593-42980593 on Build GRCh38
rs559668756 Chr.17: 42980512-42980512 on Build GRCh38
rs751337271 Chr.17: 42980549-42980549 on Build GRCh38
rs201536677 Chr.17: 42980605-42980605 on Build GRCh38
rs950629876 Chr.17: 42980493-42980493 on Build GRCh38
rs562995015 Chr.17: 42980538-42980539 on Build GRCh38
rs376633797 Chr.17: 42980603-42980603 on Build GRCh38
rs966951014 Chr.17: 42980524-42980524 on Build GRCh38
rs908498521 Chr.17: 42980515-42980515 on Build GRCh38
rs767511452 Chr.17: 42980554-42980554 on Build GRCh38
rs747361282 Chr.17: 42980582-42980582 on Build GRCh38
rs983306758 Chr.17: 42980642-42980642 on Build GRCh38
rs372920205 Chr.17: 42980589-42980589 on Build GRCh38
rs778475180 Chr.17: 42980575-42980575 on Build GRCh38
rs774457362 Chr.17: 42980601-42980601 on Build GRCh38
rs372010788 Chr.17: 42980569-42980569 on Build GRCh38
rs530398987 Chr.17: 42980595-42980595 on Build GRCh38
rs574795462 Chr.17: 42980552-42980552 on Build GRCh38
rs989122218 Chr.17: 42980501-42980501 on Build GRCh38
rs1003275151 Chr.17: 42980445-42980445 on Build GRCh38

View Full Product Details