Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7436880-7437117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM102
Entrez Gene ID
284114
Gene Name
transmembrane protein 102
Gene Aliases
CBAP
UniGene
Hs.655662
Gene Chromosome Location
Chr.17: 7435443-7437679 on Build GRCh38
Transcripts
NM_001320444.1 NM_178518.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3783103 c.981G>T p.W327C Chr.17: 7436960-7436960 on Build GRCh38
4804104 c.1138G>T p.A380S Chr.17: 7437117-7437117 on Build GRCh38
6147941 c.999C>T p.S333S Chr.17: 7436978-7436978 on Build GRCh38
2150096 c.915T>A p.A305A Chr.17: 7436894-7436894 on Build GRCh38
1735322 c.1031C>T p.S344L Chr.17: 7437010-7437010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529120197 Chr.17: 7436976-7436976 on Build GRCh38
rs758774207 Chr.17: 7436954-7436954 on Build GRCh38
rs998075673 Chr.17: 7436965-7436965 on Build GRCh38
rs983639109 Chr.17: 7437068-7437068 on Build GRCh38
rs528186177 Chr.17: 7436922-7436922 on Build GRCh38
rs775603841 Chr.17: 7437063-7437063 on Build GRCh38
rs1026438378 Chr.17: 7437070-7437070 on Build GRCh38
rs567937828 Chr.17: 7436941-7436941 on Build GRCh38
rs752073137 Chr.17: 7436979-7436979 on Build GRCh38
rs764925303 Chr.17: 7436907-7436917 on Build GRCh38
rs867215283 Chr.17: 7437033-7437033 on Build GRCh38
rs550445906 Chr.17: 7437045-7437045 on Build GRCh38
rs1030171565 Chr.17: 7436957-7436957 on Build GRCh38
rs747616570 Chr.17: 7437020-7437020 on Build GRCh38
rs755485831 Chr.17: 7436982-7436982 on Build GRCh38
rs1021553485 Chr.17: 7436916-7436916 on Build GRCh38
rs375983456 Chr.17: 7437038-7437038 on Build GRCh38
rs564203341 Chr.17: 7436913-7436913 on Build GRCh38
rs118089797 Chr.17: 7436933-7436933 on Build GRCh38
rs750683536 Chr.17: 7436953-7436953 on Build GRCh38
rs202087288 Chr.17: 7436908-7436908 on Build GRCh38
rs1007499275 Chr.17: 7437044-7437044 on Build GRCh38
rs764241700 Chr.17: 7436940-7436940 on Build GRCh38
rs765461668 Chr.17: 7437099-7437099 on Build GRCh38
rs926826375 Chr.17: 7437090-7437090 on Build GRCh38
rs777349535 Chr.17: 7437028-7437028 on Build GRCh38
rs868242394 Chr.17: 7437027-7437027 on Build GRCh38
rs772090305 Chr.17: 7437058-7437058 on Build GRCh38
rs866940466 Chr.17: 7437021-7437021 on Build GRCh38
rs769300073 Chr.17: 7437025-7437025 on Build GRCh38
rs781680285 Chr.17: 7436991-7436991 on Build GRCh38
rs918047443 Chr.17: 7437049-7437049 on Build GRCh38
rs770646047 Chr.17: 7436925-7436925 on Build GRCh38
rs772881094 Chr.17: 7436918-7436918 on Build GRCh38
rs770168038 Chr.17: 7437081-7437081 on Build GRCh38
rs868056309 Chr.17: 7436970-7436970 on Build GRCh38
rs865977940 Chr.17: 7436968-7436968 on Build GRCh38
rs970838338 Chr.17: 7437071-7437071 on Build GRCh38
rs748953825 Chr.17: 7437030-7437030 on Build GRCh38
rs111344839 Chr.17: 7436978-7436978 on Build GRCh38
rs747775232 Chr.17: 7436901-7436901 on Build GRCh38
rs759497790 Chr.17: 7437053-7437053 on Build GRCh38
rs947120003 Chr.17: 7436911-7436911 on Build GRCh38
rs759459571 Chr.17: 7436936-7436936 on Build GRCh38
rs963398807 Chr.17: 7437056-7437056 on Build GRCh38
rs200658427 Chr.17: 7436973-7436973 on Build GRCh38

View Full Product Details