Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23791711-23791944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMARCB1
Entrez Gene ID
6598
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1
Gene Aliases
BAF47, CSS3, INI1, MRD15, PPP1R144, RDT, RTPS1, SNF5, SNF5L1, SWNTS1, Sfh1p, Snr1, hSNFS
UniGene
Hs.534350
Gene Chromosome Location
Chr.22: 23786931-23834518 on Build GRCh38
Transcripts
NM_001007468.2 NM_001317946.1 NM_003073.4 XM_011530345.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1032614 c.157C>T p.R53* Chr.22: 23791819-23791819 on Build GRCh38
27946 c.132_133insTGTAC p.K45fs*12 Chr.22: 23791794-23791795 on Build GRCh38
3785510 c.226A>G p.T76A Chr.22: 23791888-23791888 on Build GRCh38
6922955 c.123delT p.S42fs*13 Chr.22: 23791785-23791785 on Build GRCh38
3939578 c.205+1G>A p.? Chr.22: 23791868-23791868 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039175631 Chr.22: 23791748-23791748 on Build GRCh38
rs587776679 Chr.22: 23791865-23791878 on Build GRCh38
rs751954991 Chr.22: 23791741-23791741 on Build GRCh38
rs779769475 Chr.22: 23791820-23791820 on Build GRCh38
rs768360679 Chr.22: 23791860-23791860 on Build GRCh38
rs774141517 Chr.22: 23791878-23791879 on Build GRCh38
rs771902726 Chr.22: 23791904-23791904 on Build GRCh38
rs748690005 Chr.22: 23791857-23791857 on Build GRCh38
rs760668060 Chr.22: 23791914-23791914 on Build GRCh38
rs373708886 Chr.22: 23791906-23791906 on Build GRCh38
rs756689642 Chr.22: 23791812-23791812 on Build GRCh38
rs370334151 Chr.22: 23791794-23791794 on Build GRCh38
rs773837831 Chr.22: 23791866-23791866 on Build GRCh38
rs746421595 Chr.22: 23791808-23791808 on Build GRCh38
rs398122368 Chr.22: 23791772-23791772 on Build GRCh38
rs387906811 Chr.22: 23791805-23791805 on Build GRCh38
rs758700723 Chr.22: 23791733-23791733 on Build GRCh38
rs939582770 Chr.22: 23791773-23791773 on Build GRCh38
rs922052006 Chr.22: 23791831-23791831 on Build GRCh38
rs747996085 Chr.22: 23791881-23791881 on Build GRCh38
rs781675982 Chr.22: 23791802-23791802 on Build GRCh38
rs377325865 Chr.22: 23791735-23791735 on Build GRCh38
rs757517233 Chr.22: 23791779-23791779 on Build GRCh38

View Full Product Details