Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 113299770-113300011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC8B1
Entrez Gene ID
80024
Gene Name
solute carrier family 8 member B1
Gene Aliases
NCKX6, NCLX, SLC24A6
UniGene
Hs.286194
Gene Chromosome Location
Chr.12: 113298766-113335142 on Build GRCh38
Transcripts
NM_024959.2 XM_006719607.2 XM_011538749.2 XM_011538750.2 XM_011538752.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
138029 c.1636C>T p.P546S Chr.12: 113299896-113299896 on Build GRCh38
430397 c.1610G>A p.S537N Chr.12: 113299922-113299922 on Build GRCh38
7174381 c.1689C>T p.F563F Chr.12: 113299843-113299843 on Build GRCh38
1161033 c.1575G>A p.L525L Chr.12: 113299957-113299957 on Build GRCh38
3359557 c.1572A>C p.G524G Chr.12: 113299960-113299960 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759052464 Chr.12: 113299814-113299814 on Build GRCh38
rs201411719 Chr.12: 113299846-113299846 on Build GRCh38
rs770343920 Chr.12: 113299806-113299806 on Build GRCh38
rs768741382 Chr.12: 113299920-113299932 on Build GRCh38
rs774113279 Chr.12: 113299809-113299809 on Build GRCh38
rs749226566 Chr.12: 113299903-113299903 on Build GRCh38
rs775229975 Chr.12: 113299869-113299870 on Build GRCh38
rs894967905 Chr.12: 113299892-113299892 on Build GRCh38
rs796879512 Chr.12: 113299856-113299856 on Build GRCh38
rs761797237 Chr.12: 113299953-113299953 on Build GRCh38
rs751541902 Chr.12: 113299936-113299936 on Build GRCh38
rs144434177 Chr.12: 113299863-113299863 on Build GRCh38
rs951791135 Chr.12: 113299866-113299866 on Build GRCh38
rs957837466 Chr.12: 113299951-113299951 on Build GRCh38
rs34833012 Chr.12: 113299918-113299918 on Build GRCh38
rs367763043 Chr.12: 113299992-113299992 on Build GRCh38
rs771670433 Chr.12: 113299917-113299917 on Build GRCh38
rs755111870 Chr.12: 113299880-113299880 on Build GRCh38
rs777653757 Chr.12: 113299977-113299977 on Build GRCh38
rs769682744 Chr.12: 113299894-113299894 on Build GRCh38
rs150854719 Chr.12: 113299824-113299824 on Build GRCh38
rs769186407 Chr.12: 113299935-113299935 on Build GRCh38
rs374615156 Chr.12: 113299962-113299962 on Build GRCh38
rs776831896 Chr.12: 113299919-113299920 on Build GRCh38
rs748511708 Chr.12: 113299963-113299963 on Build GRCh38
rs200445671 Chr.12: 113299946-113299946 on Build GRCh38
rs777927547 Chr.12: 113299901-113299901 on Build GRCh38
rs912786222 Chr.12: 113299807-113299807 on Build GRCh38
rs770491274 Chr.12: 113299908-113299908 on Build GRCh38
rs759264877 Chr.12: 113299911-113299911 on Build GRCh38
rs748009872 Chr.12: 113299893-113299893 on Build GRCh38
rs751807575 Chr.12: 113299869-113299869 on Build GRCh38
rs765571563 Chr.12: 113299921-113299921 on Build GRCh38
rs749255716 Chr.12: 113299991-113299991 on Build GRCh38
rs758560974 Chr.12: 113299858-113299858 on Build GRCh38
rs371628155 Chr.12: 113299794-113299794 on Build GRCh38
rs1054989400 Chr.12: 113299881-113299881 on Build GRCh38
rs150843854 Chr.12: 113299947-113299947 on Build GRCh38
rs931374983 Chr.12: 113299865-113299865 on Build GRCh38
rs865902233 Chr.12: 113299886-113299886 on Build GRCh38
rs773718616 Chr.12: 113299924-113299924 on Build GRCh38
rs35785542 Chr.12: 113299825-113299825 on Build GRCh38
rs747512925 Chr.12: 113299854-113299854 on Build GRCh38
rs538333342 Chr.12: 113299927-113299927 on Build GRCh38
rs758505069 Chr.12: 113299798-113299798 on Build GRCh38
rs771880330 Chr.12: 113299846-113299848 on Build GRCh38
rs747274678 Chr.12: 113299892-113299892 on Build GRCh38
rs781159760 Chr.12: 113299890-113299890 on Build GRCh38
rs745744633 Chr.12: 113299815-113299815 on Build GRCh38

View Full Product Details