Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 128806197-128806466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB7A
Entrez Gene ID
7879
Gene Name
RAB7A, member RAS oncogene family
Gene Aliases
PRO2706, RAB7
UniGene
Hs.684374
Gene Chromosome Location
Chr.3: 128726136-128814798 on Build GRCh38
Transcripts
NM_004637.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6254332 c.196G>T p.G66* Chr.3: 128806387-128806387 on Build GRCh38
1693438 c.189C>A p.D63E Chr.3: 128806380-128806380 on Build GRCh38
1037995 c.193G>A p.A65T Chr.3: 128806384-128806384 on Build GRCh38
5863438 c.197G>A p.G66E Chr.3: 128806388-128806388 on Build GRCh38
1222955 c.227C>T p.A76V Chr.3: 128806418-128806418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11549758 Chr.3: 128806393-128806393 on Build GRCh38
rs558425533 Chr.3: 128806259-128806259 on Build GRCh38
rs376642691 Chr.3: 128806374-128806374 on Build GRCh38
rs11549757 Chr.3: 128806419-128806419 on Build GRCh38
rs527920849 Chr.3: 128806280-128806280 on Build GRCh38
rs998596632 Chr.3: 128806247-128806247 on Build GRCh38
rs111584596 Chr.3: 128806285-128806285 on Build GRCh38
rs780826663 Chr.3: 128806289-128806289 on Build GRCh38
rs1025517413 Chr.3: 128806333-128806333 on Build GRCh38
rs140299645 Chr.3: 128806404-128806404 on Build GRCh38
rs191303798 Chr.3: 128806272-128806272 on Build GRCh38
rs4548 Chr.3: 128806410-128806410 on Build GRCh38
rs749968204 Chr.3: 128806372-128806372 on Build GRCh38
rs747759228 Chr.3: 128806381-128806381 on Build GRCh38
rs962910664 Chr.3: 128806320-128806320 on Build GRCh38
rs972568676 Chr.3: 128806332-128806332 on Build GRCh38
rs778176566 Chr.3: 128806443-128806443 on Build GRCh38
rs963655960 Chr.3: 128806396-128806396 on Build GRCh38
rs758757916 Chr.3: 128806422-128806422 on Build GRCh38
rs1012724147 Chr.3: 128806282-128806282 on Build GRCh38
rs1035410765 Chr.3: 128806265-128806265 on Build GRCh38

View Full Product Details