Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 235435664-235435925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBCE
Entrez Gene ID
6905
Gene Name
tubulin folding cofactor E
Gene Aliases
HRD, KCS, KCS1, pac2
UniGene
Hs.744998
Gene Chromosome Location
Chr.1: 235367360-235448968 on Build GRCh38
Transcripts
NM_001079515.2 NM_001287801.1 NM_001287802.1 NM_003193.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3485531 c.928C>T p.L310F Chr.1: 235435782-235435782 on Build GRCh38
5719052 c.775C>A p.L259I Chr.1: 235435782-235435782 on Build GRCh38
3485533 c.967C>T p.L323L Chr.1: 235435821-235435821 on Build GRCh38
4732441 c.828G>T p.L276L Chr.1: 235435835-235435835 on Build GRCh38
4732442 c.981G>T p.L327L Chr.1: 235435835-235435835 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776998898 Chr.1: 235435705-235435705 on Build GRCh38
rs759145270 Chr.1: 235435728-235435728 on Build GRCh38
rs777758117 Chr.1: 235435868-235435868 on Build GRCh38
rs959274168 Chr.1: 235435700-235435700 on Build GRCh38
rs748549375 Chr.1: 235435819-235435819 on Build GRCh38
rs773585459 Chr.1: 235435717-235435717 on Build GRCh38
rs768836850 Chr.1: 235435701-235435701 on Build GRCh38
rs751896021 Chr.1: 235435812-235435812 on Build GRCh38
rs760164430 Chr.1: 235435699-235435702 on Build GRCh38
rs185801847 Chr.1: 235435832-235435832 on Build GRCh38
rs758310255 Chr.1: 235435796-235435796 on Build GRCh38
rs200169233 Chr.1: 235435815-235435815 on Build GRCh38
rs767993266 Chr.1: 235435869-235435869 on Build GRCh38
rs757235854 Chr.1: 235435762-235435762 on Build GRCh38
rs372236046 Chr.1: 235435848-235435848 on Build GRCh38
rs764892500 Chr.1: 235435772-235435772 on Build GRCh38
rs201950450 Chr.1: 235435882-235435882 on Build GRCh38
rs750276566 Chr.1: 235435793-235435793 on Build GRCh38
rs775121475 Chr.1: 235435866-235435866 on Build GRCh38
rs370888603 Chr.1: 235435733-235435733 on Build GRCh38
rs372975302 Chr.1: 235435693-235435693 on Build GRCh38
rs927790011 Chr.1: 235435695-235435695 on Build GRCh38
rs556066006 Chr.1: 235435814-235435814 on Build GRCh38
rs763837302 Chr.1: 235435875-235435878 on Build GRCh38
rs758340135 Chr.1: 235435820-235435820 on Build GRCh38
rs770360751 Chr.1: 235435828-235435828 on Build GRCh38
rs761619502 Chr.1: 235435889-235435889 on Build GRCh38
rs769995691 Chr.1: 235435716-235435716 on Build GRCh38
rs561619540 Chr.1: 235435709-235435709 on Build GRCh38
rs375709037 Chr.1: 235435742-235435742 on Build GRCh38
rs763729563 Chr.1: 235435746-235435746 on Build GRCh38
rs780407960 Chr.1: 235435801-235435801 on Build GRCh38
rs984195019 Chr.1: 235435851-235435851 on Build GRCh38
rs775170535 Chr.1: 235435739-235435739 on Build GRCh38
rs778984741 Chr.1: 235435704-235435704 on Build GRCh38
rs368360310 Chr.1: 235435755-235435755 on Build GRCh38
rs565869709 Chr.1: 235435847-235435847 on Build GRCh38

View Full Product Details