Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12014671-12014920 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101928464
Entrez Gene ID
101928464
Gene Name
uncharacterized LOC101928464
Gene Aliases
-
UniGene
Hs.631643
Gene Chromosome Location
Chr.19: 11987617-12046275 on Build GRCh38
Transcripts
NR_134927.1 NR_134928.1 NR_134929.1 NM_001080411.2 NM_001308346.1 NM_001308348.1 NM_001308351.1 NM_001308355.1 NM_001308357.1 XM_006722661.3 XM_011527725.2 XM_011527728.2 XM_011527729.2 XM_017026413.1

Gene Symbol
ZNF433
Entrez Gene ID
163059
Gene Name
zinc finger protein 433
Gene Aliases
-
UniGene
Hs.667776
Gene Chromosome Location
Chr.19: 12014717-12035741 on Build GRCh38
Transcripts
NR_134927.1 NR_134928.1 NR_134929.1 NM_001080411.2 NM_001308346.1 NM_001308348.1 NM_001308351.1 NM_001308355.1 NM_001308357.1 XM_006722661.3 XM_011527725.2 XM_011527728.2 XM_011527729.2 XM_017026413.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5570246 c.2015C>T p.T672I Chr.19: 12014852-12014852 on Build GRCh38
4428815 c.2008_2009insA p.I670fs*>5 Chr.19: 12014858-12014859 on Build GRCh38
7179387 c.1967G>A p.R656K Chr.19: 12014900-12014900 on Build GRCh38
4140257 c.2013C>T p.D671D Chr.19: 12014854-12014854 on Build GRCh38
238485 c.1959A>G p.K653K Chr.19: 12014908-12014908 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764900725 Chr.19: 12014855-12014855 on Build GRCh38
rs763989599 Chr.19: 12014837-12014837 on Build GRCh38
rs753823177 Chr.19: 12014838-12014838 on Build GRCh38
rs969810273 Chr.19: 12014816-12014816 on Build GRCh38
rs900969774 Chr.19: 12014706-12014706 on Build GRCh38
rs190101419 Chr.19: 12014856-12014856 on Build GRCh38
rs776890476 Chr.19: 12014826-12014834 on Build GRCh38
rs1023150332 Chr.19: 12014794-12014794 on Build GRCh38
rs753047612 Chr.19: 12014730-12014730 on Build GRCh38
rs891058422 Chr.19: 12014698-12014698 on Build GRCh38
rs191482684 Chr.19: 12014766-12014766 on Build GRCh38
rs558794972 Chr.19: 12014753-12014753 on Build GRCh38
rs1020786943 Chr.19: 12014697-12014697 on Build GRCh38
rs771245238 Chr.19: 12014804-12014804 on Build GRCh38
rs376882241 Chr.19: 12014817-12014817 on Build GRCh38
rs760813319 Chr.19: 12014834-12014834 on Build GRCh38
rs746511641 Chr.19: 12014880-12014880 on Build GRCh38
rs537520980 Chr.19: 12014728-12014728 on Build GRCh38
rs750236446 Chr.19: 12014863-12014863 on Build GRCh38
rs764567045 Chr.19: 12014735-12014736 on Build GRCh38
rs185252567 Chr.19: 12014790-12014790 on Build GRCh38
rs989018251 Chr.19: 12014851-12014851 on Build GRCh38
rs774797058 Chr.19: 12014815-12014815 on Build GRCh38
rs559559698 Chr.19: 12014769-12014769 on Build GRCh38
rs775537208 Chr.19: 12014831-12014831 on Build GRCh38
rs779908822 Chr.19: 12014875-12014875 on Build GRCh38
rs371642190 Chr.19: 12014883-12014883 on Build GRCh38
rs576380559 Chr.19: 12014708-12014708 on Build GRCh38
rs367705892 Chr.19: 12014717-12014719 on Build GRCh38
rs187041182 Chr.19: 12014765-12014765 on Build GRCh38
rs953778378 Chr.19: 12014771-12014771 on Build GRCh38
rs1004126248 Chr.19: 12014736-12014739 on Build GRCh38
rs767797905 Chr.19: 12014820-12014820 on Build GRCh38
rs367917580 Chr.19: 12014869-12014869 on Build GRCh38
rs934946792 Chr.19: 12014840-12014840 on Build GRCh38
rs917436434 Chr.19: 12014841-12014841 on Build GRCh38
rs757241745 Chr.19: 12014852-12014852 on Build GRCh38
rs983784704 Chr.19: 12014774-12014774 on Build GRCh38

View Full Product Details