Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 135359911-135360170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF449
Entrez Gene ID
203523
Gene Name
zinc finger protein 449
Gene Aliases
ZSCAN19
UniGene
Hs.28780
Gene Chromosome Location
Chr.X: 135344098-135363413 on Build GRCh38
Transcripts
NM_152695.5 XM_011531312.2 XM_011531313.2 XM_011531314.2 XM_017029350.1 XM_017029351.1 XM_017029352.1

SNP Detail

SNP ID Chromosome Location
rs950709585 Chr.X: 135360110-135360110 on Build GRCh38
rs782722257 Chr.X: 135360013-135360013 on Build GRCh38
rs782681513 Chr.X: 135360036-135360037 on Build GRCh38
rs782282817 Chr.X: 135360044-135360044 on Build GRCh38
rs782758673 Chr.X: 135360036-135360036 on Build GRCh38
rs782254149 Chr.X: 135359947-135359947 on Build GRCh38
rs782158487 Chr.X: 135360033-135360034 on Build GRCh38
rs1026189937 Chr.X: 135360030-135360030 on Build GRCh38
rs782026565 Chr.X: 135359985-135359985 on Build GRCh38
rs781869775 Chr.X: 135360037-135360037 on Build GRCh38
rs782397742 Chr.X: 135360117-135360117 on Build GRCh38
rs782807080 Chr.X: 135360037-135360037 on Build GRCh38
rs782803029 Chr.X: 135360048-135360048 on Build GRCh38
rs370151328 Chr.X: 135360027-135360027 on Build GRCh38
rs781950225 Chr.X: 135360023-135360023 on Build GRCh38
rs782039605 Chr.X: 135360011-135360011 on Build GRCh38
rs782394485 Chr.X: 135359977-135359977 on Build GRCh38

View Full Product Details