Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 27101254-27101495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXA2
Entrez Gene ID
3199
Gene Name
homeobox A2
Gene Aliases
HOX1K, MCOHI
UniGene
Hs.445239
Gene Chromosome Location
Chr.7: 27100354-27102775 on Build GRCh38
Transcripts
NM_006735.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3027088 c.507C>A p.C169* Chr.7: 27101350-27101350 on Build GRCh38
7096640 c.431G>A p.R144Q Chr.7: 27101426-27101426 on Build GRCh38
107066 c.519G>A p.R173R Chr.7: 27101338-27101338 on Build GRCh38
3880388 c.420C>T p.G140G Chr.7: 27101437-27101437 on Build GRCh38
296298 c.432G>T p.R144R Chr.7: 27101425-27101425 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757201406 Chr.7: 27101408-27101408 on Build GRCh38
rs752989766 Chr.7: 27101356-27101356 on Build GRCh38
rs755775462 Chr.7: 27101432-27101432 on Build GRCh38
rs762796761 Chr.7: 27101359-27101359 on Build GRCh38
rs887991299 Chr.7: 27101324-27101324 on Build GRCh38
rs780752179 Chr.7: 27101417-27101417 on Build GRCh38
rs763986379 Chr.7: 27101365-27101365 on Build GRCh38
rs779221330 Chr.7: 27101275-27101275 on Build GRCh38
rs771410610 Chr.7: 27101445-27101445 on Build GRCh38
rs748564970 Chr.7: 27101303-27101303 on Build GRCh38
rs119489104 Chr.7: 27101301-27101301 on Build GRCh38
rs772381717 Chr.7: 27101438-27101438 on Build GRCh38
rs772364626 Chr.7: 27101439-27101439 on Build GRCh38
rs769998504 Chr.7: 27101468-27101468 on Build GRCh38
rs761096280 Chr.7: 27101323-27101323 on Build GRCh38
rs140982794 Chr.7: 27101338-27101338 on Build GRCh38
rs749965554 Chr.7: 27101426-27101426 on Build GRCh38
rs139504171 Chr.7: 27101463-27101463 on Build GRCh38
rs1011085616 Chr.7: 27101431-27101431 on Build GRCh38
rs773638633 Chr.7: 27101320-27101320 on Build GRCh38
rs751457659 Chr.7: 27101395-27101395 on Build GRCh38
rs776638973 Chr.7: 27101336-27101336 on Build GRCh38
rs748975641 Chr.7: 27101437-27101437 on Build GRCh38
rs779619613 Chr.7: 27101436-27101436 on Build GRCh38
rs765603928 Chr.7: 27101344-27101344 on Build GRCh38
rs747522109 Chr.7: 27101440-27101440 on Build GRCh38
rs776760448 Chr.7: 27101446-27101446 on Build GRCh38

View Full Product Details