Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 23703640-23703899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATAD2B
Entrez Gene ID
54454
Gene Name
ATPase family, AAA domain containing 2B
Gene Aliases
-
UniGene
Hs.467862
Gene Chromosome Location
Chr.2: 23689710-23927114 on Build GRCh38
Transcripts
XM_006712030.3 NM_052920.1 XM_006711929.3 XM_011532501.2 XM_017003264.1

Gene Symbol
KLHL29
Entrez Gene ID
114818
Gene Name
kelch like family member 29
Gene Aliases
KBTBD9
UniGene
Hs.130593
Gene Chromosome Location
Chr.2: 23385419-23708613 on Build GRCh38
Transcripts
XM_006712030.3 NM_052920.1 XM_006711929.3 XM_011532501.2 XM_017003264.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5734890 c.2315C>G p.P772R Chr.2: 23703734-23703734 on Build GRCh38
7291526 c.1688G>A p.G563D Chr.2: 23703770-23703770 on Build GRCh38
5630758 c.1719C>G p.Y573* Chr.2: 23703801-23703801 on Build GRCh38
5396597 c.1781+1G>A p.? Chr.2: 23703864-23703864 on Build GRCh38
6224665 c.1690G>A p.G564R Chr.2: 23703772-23703772 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs945365272 Chr.2: 23703740-23703740 on Build GRCh38
rs989478689 Chr.2: 23703675-23703675 on Build GRCh38
rs952320454 Chr.2: 23703877-23703877 on Build GRCh38
rs3795948 Chr.2: 23703789-23703789 on Build GRCh38
rs1045478508 Chr.2: 23703851-23703851 on Build GRCh38
rs376299706 Chr.2: 23703689-23703689 on Build GRCh38
rs200173076 Chr.2: 23703678-23703678 on Build GRCh38
rs758006127 Chr.2: 23703706-23703706 on Build GRCh38
rs932096623 Chr.2: 23703685-23703685 on Build GRCh38
rs189934640 Chr.2: 23703871-23703871 on Build GRCh38
rs748058181 Chr.2: 23703815-23703815 on Build GRCh38
rs554907955 Chr.2: 23703771-23703771 on Build GRCh38
rs1013906186 Chr.2: 23703866-23703866 on Build GRCh38
rs749905554 Chr.2: 23703704-23703704 on Build GRCh38
rs574075853 Chr.2: 23703805-23703805 on Build GRCh38
rs775788027 Chr.2: 23703879-23703879 on Build GRCh38
rs911939234 Chr.2: 23703694-23703694 on Build GRCh38
rs1027770618 Chr.2: 23703876-23703876 on Build GRCh38
rs768204340 Chr.2: 23703744-23703744 on Build GRCh38
rs778254329 Chr.2: 23703690-23703690 on Build GRCh38
rs368696669 Chr.2: 23703801-23703801 on Build GRCh38
rs894161293 Chr.2: 23703837-23703837 on Build GRCh38
rs759612469 Chr.2: 23703875-23703875 on Build GRCh38
rs925890595 Chr.2: 23703749-23703749 on Build GRCh38
rs986256682 Chr.2: 23703666-23703666 on Build GRCh38
rs772197706 Chr.2: 23703878-23703878 on Build GRCh38
rs772009025 Chr.2: 23703827-23703827 on Build GRCh38
rs966085386 Chr.2: 23703697-23703697 on Build GRCh38
rs185152824 Chr.2: 23703757-23703757 on Build GRCh38
rs371026611 Chr.2: 23703701-23703701 on Build GRCh38
rs924225817 Chr.2: 23703664-23703664 on Build GRCh38
rs1044121142 Chr.2: 23703743-23703743 on Build GRCh38

View Full Product Details