Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 2250806-2251077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUDT1
Entrez Gene ID
4521
Gene Name
nudix hydrolase 1
Gene Aliases
MTH1
UniGene
Hs.534331
Gene Chromosome Location
Chr.7: 2242222-2251145 on Build GRCh38
Transcripts
NM_002452.3 NM_198948.1 NM_198949.1 NM_198950.1 NM_198952.1 NM_198953.1 NM_198954.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5592182 c.422C>T p.P141L Chr.7: 2250883-2250883 on Build GRCh38
6731119 c.522C>T p.R174R Chr.7: 2250983-2250983 on Build GRCh38
6731120 c.374G>A p.R125H Chr.7: 2250835-2250835 on Build GRCh38
1237871 c.496G>A p.D166N Chr.7: 2250957-2250957 on Build GRCh38
6731121 c.384G>A p.W128* Chr.7: 2250845-2250845 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747740732 Chr.7: 2250920-2250920 on Build GRCh38
rs567724444 Chr.7: 2250850-2250850 on Build GRCh38
rs780761617 Chr.7: 2251045-2251045 on Build GRCh38
rs377540407 Chr.7: 2250890-2250890 on Build GRCh38
rs756663427 Chr.7: 2251041-2251041 on Build GRCh38
rs142532068 Chr.7: 2250966-2250966 on Build GRCh38
rs766938119 Chr.7: 2251005-2251006 on Build GRCh38
rs577899382 Chr.7: 2250967-2250967 on Build GRCh38
rs746332503 Chr.7: 2251002-2251002 on Build GRCh38
rs202044583 Chr.7: 2250835-2250835 on Build GRCh38
rs774269541 Chr.7: 2250971-2250971 on Build GRCh38
rs771501296 Chr.7: 2250849-2250849 on Build GRCh38
rs763276275 Chr.7: 2250880-2250880 on Build GRCh38
rs753038773 Chr.7: 2250877-2250877 on Build GRCh38
rs146088002 Chr.7: 2250925-2250925 on Build GRCh38
rs778683326 Chr.7: 2250911-2250911 on Build GRCh38
rs764448314 Chr.7: 2250969-2250969 on Build GRCh38
rs138059598 Chr.7: 2250947-2250947 on Build GRCh38
rs1040303238 Chr.7: 2250941-2250941 on Build GRCh38
rs761039063 Chr.7: 2250848-2250848 on Build GRCh38
rs143631820 Chr.7: 2250884-2250884 on Build GRCh38
rs761944519 Chr.7: 2251018-2251018 on Build GRCh38
rs114380492 Chr.7: 2250982-2250982 on Build GRCh38
rs753559188 Chr.7: 2250897-2250897 on Build GRCh38
rs35573651 Chr.7: 2251037-2251037 on Build GRCh38
rs200747756 Chr.7: 2250973-2250973 on Build GRCh38
rs768169199 Chr.7: 2250828-2250828 on Build GRCh38
rs754832477 Chr.7: 2250898-2250898 on Build GRCh38
rs772723311 Chr.7: 2250842-2250842 on Build GRCh38
rs971522766 Chr.7: 2250896-2250896 on Build GRCh38
rs141118084 Chr.7: 2251033-2251033 on Build GRCh38
rs149666869 Chr.7: 2251003-2251003 on Build GRCh38
rs770220369 Chr.7: 2250957-2250957 on Build GRCh38
rs747651511 Chr.7: 2250834-2250834 on Build GRCh38
rs139949371 Chr.7: 2250984-2250984 on Build GRCh38
rs773187512 Chr.7: 2251019-2251019 on Build GRCh38
rs759996258 Chr.7: 2250861-2250861 on Build GRCh38
rs182662727 Chr.7: 2250933-2250933 on Build GRCh38
rs766221010 Chr.7: 2250977-2250977 on Build GRCh38
rs749716688 Chr.7: 2251012-2251012 on Build GRCh38
rs768984899 Chr.7: 2251013-2251013 on Build GRCh38
rs764217206 Chr.7: 2250881-2250881 on Build GRCh38
rs772860020 Chr.7: 2251011-2251011 on Build GRCh38
rs765546387 Chr.7: 2250876-2250876 on Build GRCh38
rs768762199 Chr.7: 2250916-2250918 on Build GRCh38
rs141913410 Chr.7: 2250888-2250888 on Build GRCh38
rs372938387 Chr.7: 2251029-2251029 on Build GRCh38
rs148931821 Chr.7: 2250831-2250831 on Build GRCh38
rs763653029 Chr.7: 2251031-2251031 on Build GRCh38
rs780576043 Chr.7: 2251001-2251001 on Build GRCh38
rs547745300 Chr.7: 2251059-2251059 on Build GRCh38
rs573728470 Chr.7: 2250995-2250995 on Build GRCh38
rs538686319 Chr.7: 2250932-2250932 on Build GRCh38
rs777532626 Chr.7: 2250989-2250989 on Build GRCh38
rs1799832 Chr.7: 2250887-2250887 on Build GRCh38
rs752468162 Chr.7: 2251028-2251028 on Build GRCh38
rs1000625228 Chr.7: 2250838-2250838 on Build GRCh38
rs529226594 Chr.7: 2251008-2251008 on Build GRCh38
rs774699263 Chr.7: 2251017-2251017 on Build GRCh38
rs760299497 Chr.7: 2251018-2251018 on Build GRCh38
rs759429398 Chr.7: 2251025-2251025 on Build GRCh38
rs752521039 Chr.7: 2250983-2250983 on Build GRCh38
rs1042577663 Chr.7: 2250971-2250972 on Build GRCh38
rs774450732 Chr.7: 2250936-2250937 on Build GRCh38
rs753740991 Chr.7: 2250978-2250978 on Build GRCh38
rs765200874 Chr.7: 2251027-2251027 on Build GRCh38
rs372216748 Chr.7: 2251051-2251051 on Build GRCh38
rs754921444 Chr.7: 2250981-2250981 on Build GRCh38
rs756878176 Chr.7: 2251034-2251034 on Build GRCh38
rs761344946 Chr.7: 2250976-2250976 on Build GRCh38
rs746490760 Chr.7: 2250994-2250994 on Build GRCh38
rs749663700 Chr.7: 2251047-2251047 on Build GRCh38
rs1062492 Chr.7: 2251050-2251050 on Build GRCh38

View Full Product Details