Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 70145277-70145550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TYSND1
Entrez Gene ID
219743
Gene Name
trypsin domain containing 1
Gene Aliases
NET41
UniGene
Hs.533655
Gene Chromosome Location
Chr.10: 70137981-70146740 on Build GRCh38
Transcripts
NM_001040273.2 NM_173555.3 NR_073580.1 NR_073581.1 NR_073582.1 NR_073591.1 NR_073592.1 NR_073593.1 NR_073594.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2149334 c.1136C>T p.A379V Chr.10: 70145451-70145451 on Build GRCh38
6269779 c.1155C>A p.S385S Chr.10: 70145432-70145432 on Build GRCh38
4738551 c.1107C>A p.T369T Chr.10: 70145480-70145480 on Build GRCh38
7332815 c.1113G>T p.R371R Chr.10: 70145474-70145474 on Build GRCh38
4015473 c.1058G>A p.G353D Chr.10: 70145529-70145529 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541825991 Chr.10: 70145454-70145454 on Build GRCh38
rs765801570 Chr.10: 70145517-70145517 on Build GRCh38
rs922730257 Chr.10: 70145339-70145339 on Build GRCh38
rs981094829 Chr.10: 70145321-70145321 on Build GRCh38
rs767205352 Chr.10: 70145436-70145436 on Build GRCh38
rs773180600 Chr.10: 70145413-70145413 on Build GRCh38
rs779983570 Chr.10: 70145436-70145439 on Build GRCh38
rs200098587 Chr.10: 70145376-70145376 on Build GRCh38
rs752360175 Chr.10: 70145449-70145449 on Build GRCh38
rs373686566 Chr.10: 70145482-70145482 on Build GRCh38
rs779227788 Chr.10: 70145466-70145466 on Build GRCh38
rs779308906 Chr.10: 70145383-70145383 on Build GRCh38
rs376874341 Chr.10: 70145531-70145531 on Build GRCh38
rs934161942 Chr.10: 70145349-70145350 on Build GRCh38
rs932791242 Chr.10: 70145397-70145397 on Build GRCh38
rs770699328 Chr.10: 70145422-70145422 on Build GRCh38
rs770914763 Chr.10: 70145497-70145497 on Build GRCh38
rs944285595 Chr.10: 70145465-70145465 on Build GRCh38
rs560647209 Chr.10: 70145476-70145476 on Build GRCh38
rs745708882 Chr.10: 70145499-70145499 on Build GRCh38
rs920110824 Chr.10: 70145394-70145394 on Build GRCh38
rs376923738 Chr.10: 70145398-70145398 on Build GRCh38
rs776291108 Chr.10: 70145514-70145514 on Build GRCh38
rs572405383 Chr.10: 70145515-70145515 on Build GRCh38
rs908205235 Chr.10: 70145329-70145329 on Build GRCh38
rs899077659 Chr.10: 70145426-70145426 on Build GRCh38
rs749281400 Chr.10: 70145496-70145496 on Build GRCh38
rs759195775 Chr.10: 70145429-70145429 on Build GRCh38
rs780343652 Chr.10: 70145475-70145475 on Build GRCh38
rs760743439 Chr.10: 70145385-70145385 on Build GRCh38
rs574800270 Chr.10: 70145450-70145450 on Build GRCh38
rs115327224 Chr.10: 70145402-70145402 on Build GRCh38
rs750844425 Chr.10: 70145530-70145530 on Build GRCh38
rs149277739 Chr.10: 70145502-70145503 on Build GRCh38
rs764220845 Chr.10: 70145416-70145416 on Build GRCh38
rs763679228 Chr.10: 70145451-70145451 on Build GRCh38
rs1022861876 Chr.10: 70145303-70145303 on Build GRCh38
rs912774302 Chr.10: 70145427-70145427 on Build GRCh38
rs961180528 Chr.10: 70145354-70145354 on Build GRCh38
rs750801224 Chr.10: 70145467-70145467 on Build GRCh38
rs760376665 Chr.10: 70145506-70145506 on Build GRCh38
rs199952796 Chr.10: 70145502-70145502 on Build GRCh38
rs757765472 Chr.10: 70145461-70145461 on Build GRCh38
rs758648017 Chr.10: 70145471-70145471 on Build GRCh38
rs763767465 Chr.10: 70145511-70145511 on Build GRCh38
rs755089159 Chr.10: 70145480-70145480 on Build GRCh38
rs576765554 Chr.10: 70145307-70145307 on Build GRCh38
rs867292004 Chr.10: 70145524-70145524 on Build GRCh38
rs774183123 Chr.10: 70145428-70145428 on Build GRCh38
rs768671020 Chr.10: 70145411-70145411 on Build GRCh38

View Full Product Details