Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 264784-265057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACP1
Entrez Gene ID
52
Gene Name
acid phosphatase 1, soluble
Gene Aliases
HAAP, LMW-PTP
UniGene
Hs.558296
Gene Chromosome Location
Chr.2: 264869-278283 on Build GRCh38
Transcripts
NM_001040649.2 NM_004300.3 NM_007099.3 NR_024080.1 NM_001282682.1 NR_104224.1

Gene Symbol
SH3YL1
Entrez Gene ID
26751
Gene Name
SH3 and SYLF domain containing 1
Gene Aliases
RAY
UniGene
Hs.515951
Gene Chromosome Location
Chr.2: 218136-264866 on Build GRCh38
Transcripts
NM_001040649.2 NM_004300.3 NM_007099.3 NR_024080.1 NM_001282682.1 NR_104224.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6592420 c.18_19insA p.S8fs*9 Chr.2: 264982-264983 on Build GRCh38
3364687 c.40C>G p.L14V Chr.2: 265004-265004 on Build GRCh38
6592415 c.11A>T p.Q4L Chr.2: 264975-264975 on Build GRCh38
242781 c.18C>G p.T6T Chr.2: 264982-264982 on Build GRCh38
7227614 c.14C>T p.A5V Chr.2: 264978-264978 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs986406946 Chr.2: 264822-264822 on Build GRCh38
rs767660291 Chr.2: 265016-265016 on Build GRCh38
rs770786313 Chr.2: 264946-264946 on Build GRCh38
rs1029234934 Chr.2: 264802-264802 on Build GRCh38
rs778712454 Chr.2: 265019-265019 on Build GRCh38
rs774092290 Chr.2: 264923-264923 on Build GRCh38
rs752826543 Chr.2: 264944-264945 on Build GRCh38
rs111341344 Chr.2: 264879-264879 on Build GRCh38
rs145643708 Chr.2: 264965-264965 on Build GRCh38
rs1043137136 Chr.2: 264900-264902 on Build GRCh38
rs938510212 Chr.2: 264882-264882 on Build GRCh38
rs201289787 Chr.2: 265010-265010 on Build GRCh38
rs995796859 Chr.2: 264898-264898 on Build GRCh38
rs754275656 Chr.2: 264971-264971 on Build GRCh38
rs763210317 Chr.2: 265013-265013 on Build GRCh38
rs997967890 Chr.2: 265026-265026 on Build GRCh38
rs1044159971 Chr.2: 264875-264875 on Build GRCh38
rs771602723 Chr.2: 264958-264958 on Build GRCh38
rs764554243 Chr.2: 264933-264933 on Build GRCh38
rs778962336 Chr.2: 264983-264983 on Build GRCh38
rs755699886 Chr.2: 264932-264932 on Build GRCh38
rs762338324 Chr.2: 264972-264972 on Build GRCh38
rs367983442 Chr.2: 264913-264913 on Build GRCh38
rs780131557 Chr.2: 264979-264979 on Build GRCh38
rs764998912 Chr.2: 265006-265006 on Build GRCh38
rs754821888 Chr.2: 264832-264832 on Build GRCh38
rs901932466 Chr.2: 265024-265024 on Build GRCh38
rs989838811 Chr.2: 264856-264856 on Build GRCh38
rs775006639 Chr.2: 264918-264924 on Build GRCh38
rs376349932 Chr.2: 265036-265036 on Build GRCh38
rs573062193 Chr.2: 264823-264823 on Build GRCh38
rs761003573 Chr.2: 264976-264976 on Build GRCh38
rs768112455 Chr.2: 264989-264989 on Build GRCh38
rs767662864 Chr.2: 264943-264943 on Build GRCh38
rs372441052 Chr.2: 264941-264941 on Build GRCh38
rs147831354 Chr.2: 264982-264982 on Build GRCh38
rs534121767 Chr.2: 264833-264833 on Build GRCh38
rs774993891 Chr.2: 264988-264988 on Build GRCh38
rs568470887 Chr.2: 265003-265003 on Build GRCh38
rs140935600 Chr.2: 264975-264975 on Build GRCh38
rs759478140 Chr.2: 264932-264932 on Build GRCh38
rs919712463 Chr.2: 264861-264861 on Build GRCh38
rs202179100 Chr.2: 264981-264981 on Build GRCh38
rs147428558 Chr.2: 265004-265004 on Build GRCh38
rs766776227 Chr.2: 265014-265014 on Build GRCh38
rs769828590 Chr.2: 264914-264914 on Build GRCh38
rs376020864 Chr.2: 265020-265020 on Build GRCh38
rs371845175 Chr.2: 264916-264916 on Build GRCh38
rs556073250 Chr.2: 264844-264844 on Build GRCh38
rs11691572 Chr.2: 264985-264985 on Build GRCh38
rs374297884 Chr.2: 265021-265021 on Build GRCh38
rs765630438 Chr.2: 265005-265005 on Build GRCh38
rs116051772 Chr.2: 265015-265015 on Build GRCh38
rs765536704 Chr.2: 264976-264976 on Build GRCh38
rs557935748 Chr.2: 264814-264814 on Build GRCh38
rs752174815 Chr.2: 264929-264929 on Build GRCh38
rs768919795 Chr.2: 264943-264943 on Build GRCh38
rs762834566 Chr.2: 264918-264918 on Build GRCh38
rs918377642 Chr.2: 265008-265008 on Build GRCh38
rs759772284 Chr.2: 264846-264846 on Build GRCh38
rs887778647 Chr.2: 264910-264910 on Build GRCh38
rs376391902 Chr.2: 264930-264930 on Build GRCh38
rs1049054480 Chr.2: 264908-264908 on Build GRCh38
rs11696071 Chr.2: 264948-264948 on Build GRCh38
rs539121873 Chr.2: 264806-264806 on Build GRCh38
rs759184328 Chr.2: 264954-264954 on Build GRCh38
rs773318542 Chr.2: 264917-264917 on Build GRCh38
rs938591578 Chr.2: 264886-264886 on Build GRCh38
rs200105013 Chr.2: 264937-264937 on Build GRCh38
rs746614764 Chr.2: 265034-265034 on Build GRCh38
rs143584646 Chr.2: 265017-265017 on Build GRCh38
rs946016572 Chr.2: 264986-264986 on Build GRCh38
rs369916711 Chr.2: 264978-264978 on Build GRCh38
rs4854300 Chr.2: 264895-264895 on Build GRCh38
rs1032221395 Chr.2: 264957-264957 on Build GRCh38
rs972408605 Chr.2: 264950-264950 on Build GRCh38
rs369693627 Chr.2: 264935-264935 on Build GRCh38
rs767017144 Chr.2: 264928-264928 on Build GRCh38
rs1031876142 Chr.2: 265031-265031 on Build GRCh38
rs763654173 Chr.2: 264969-264969 on Build GRCh38
rs369444515 Chr.2: 264980-264980 on Build GRCh38
rs562703679 Chr.2: 264890-264890 on Build GRCh38
rs972461994 Chr.2: 264853-264853 on Build GRCh38
rs766332062 Chr.2: 264922-264922 on Build GRCh38
rs932071954 Chr.2: 265011-265011 on Build GRCh38
rs780372162 Chr.2: 264939-264939 on Build GRCh38
rs574229810 Chr.2: 264862-264862 on Build GRCh38
rs914199776 Chr.2: 264857-264857 on Build GRCh38
rs754410823 Chr.2: 264934-264934 on Build GRCh38
rs528331795 Chr.2: 264959-264959 on Build GRCh38

View Full Product Details