Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 16690304-16690569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC144A
Entrez Gene ID
9720
Gene Name
coiled-coil domain containing 144A
Gene Aliases
-
UniGene
Hs.745246
Gene Chromosome Location
Chr.17: 16666768-16774999 on Build GRCh38
Transcripts
NM_014695.2 NR_130142.1 XM_011524089.2 XM_017025429.1 XM_017025430.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3514641 c.72G>A p.R24R Chr.17: 16690472-16690472 on Build GRCh38
6621647 c.124G>A p.G42R Chr.17: 16690524-16690524 on Build GRCh38
3514642 c.72G>A p.R24R Chr.17: 16690472-16690472 on Build GRCh38
4556685 c.69G>A p.T23T Chr.17: 16690469-16690469 on Build GRCh38
6287731 c.46C>G p.P16A Chr.17: 16690446-16690446 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763956623 Chr.17: 16690380-16690380 on Build GRCh38
rs772302260 Chr.17: 16690454-16690454 on Build GRCh38
rs748367093 Chr.17: 16690496-16690522 on Build GRCh38
rs755043170 Chr.17: 16690365-16690366 on Build GRCh38
rs757757152 Chr.17: 16690500-16690500 on Build GRCh38
rs759229691 Chr.17: 16690541-16690541 on Build GRCh38
rs776670944 Chr.17: 16690522-16690522 on Build GRCh38
rs2928658 Chr.17: 16690405-16690405 on Build GRCh38
rs750183409 Chr.17: 16690480-16690480 on Build GRCh38
rs769731047 Chr.17: 16690529-16690529 on Build GRCh38
rs779324033 Chr.17: 16690502-16690502 on Build GRCh38
rs562771471 Chr.17: 16690540-16690540 on Build GRCh38
rs756328470 Chr.17: 16690506-16690507 on Build GRCh38
rs751268620 Chr.17: 16690485-16690485 on Build GRCh38
rs557850602 Chr.17: 16690423-16690423 on Build GRCh38
rs758047500 Chr.17: 16690401-16690401 on Build GRCh38
rs749029167 Chr.17: 16690534-16690540 on Build GRCh38
rs781317826 Chr.17: 16690438-16690439 on Build GRCh38
rs771404713 Chr.17: 16690426-16690426 on Build GRCh38
rs780843051 Chr.17: 16690495-16690495 on Build GRCh38
rs367735394 Chr.17: 16690418-16690418 on Build GRCh38
rs879173110 Chr.17: 16690323-16690323 on Build GRCh38
rs773026244 Chr.17: 16690532-16690532 on Build GRCh38
rs758157607 Chr.17: 16690481-16690481 on Build GRCh38
rs746181106 Chr.17: 16690503-16690503 on Build GRCh38
rs760691655 Chr.17: 16690457-16690457 on Build GRCh38
rs577983126 Chr.17: 16690425-16690425 on Build GRCh38
rs201938569 Chr.17: 16690361-16690361 on Build GRCh38
rs2349265 Chr.17: 16690484-16690484 on Build GRCh38
rs777460449 Chr.17: 16690514-16690540 on Build GRCh38
rs774742134 Chr.17: 16690440-16690440 on Build GRCh38
rs772340627 Chr.17: 16690505-16690505 on Build GRCh38
rs750226568 Chr.17: 16690398-16690398 on Build GRCh38
rs201730519 Chr.17: 16690547-16690547 on Build GRCh38
rs1025575242 Chr.17: 16690330-16690330 on Build GRCh38
rs765136331 Chr.17: 16690479-16690479 on Build GRCh38
rs747229684 Chr.17: 16690516-16690516 on Build GRCh38
rs759795875 Chr.17: 16690351-16690351 on Build GRCh38
rs200445302 Chr.17: 16690513-16690513 on Build GRCh38
rs764147507 Chr.17: 16690458-16690458 on Build GRCh38
rs779422079 Chr.17: 16690548-16690548 on Build GRCh38
rs895404347 Chr.17: 16690326-16690326 on Build GRCh38
rs574092546 Chr.17: 16690524-16690524 on Build GRCh38
rs775583093 Chr.17: 16690455-16690455 on Build GRCh38
rs2928657 Chr.17: 16690488-16690488 on Build GRCh38
rs760778514 Chr.17: 16690368-16690368 on Build GRCh38
rs528522303 Chr.17: 16690332-16690332 on Build GRCh38
rs982973555 Chr.17: 16690446-16690446 on Build GRCh38
rs542811265 Chr.17: 16690537-16690537 on Build GRCh38
rs200340971 Chr.17: 16690381-16690381 on Build GRCh38
rs201250638 Chr.17: 16690519-16690519 on Build GRCh38
rs767669501 Chr.17: 16690359-16690359 on Build GRCh38
rs765077555 Chr.17: 16690397-16690397 on Build GRCh38
rs751704438 Chr.17: 16690357-16690361 on Build GRCh38
rs895370300 Chr.17: 16690476-16690476 on Build GRCh38
rs761708441 Chr.17: 16690477-16690477 on Build GRCh38
rs370924631 Chr.17: 16690411-16690411 on Build GRCh38
rs906604285 Chr.17: 16690344-16690344 on Build GRCh38
rs757244926 Chr.17: 16690386-16690386 on Build GRCh38
rs368437733 Chr.17: 16690538-16690538 on Build GRCh38
rs371265519 Chr.17: 16690453-16690453 on Build GRCh38
rs747697017 Chr.17: 16690498-16690498 on Build GRCh38
rs199720294 Chr.17: 16690510-16690510 on Build GRCh38
rs776760724 Chr.17: 16690465-16690465 on Build GRCh38

View Full Product Details