Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 109624539-109624766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNE5
Entrez Gene ID
23630
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 5
Gene Aliases
KCNE1L
UniGene
Hs.522753
Gene Chromosome Location
Chr.X: 109623700-109625164 on Build GRCh38
Transcripts
NM_012282.2

SNP Detail

SNP ID Chromosome Location
rs988966138 Chr.X: 109624747-109624747 on Build GRCh38
rs775988479 Chr.X: 109624682-109624682 on Build GRCh38
rs61729624 Chr.X: 109624744-109624744 on Build GRCh38
rs769195237 Chr.X: 109624735-109624735 on Build GRCh38
rs750566339 Chr.X: 109624703-109624703 on Build GRCh38
rs1047427572 Chr.X: 109624611-109624611 on Build GRCh38
rs776874809 Chr.X: 109624624-109624624 on Build GRCh38
rs765270209 Chr.X: 109624718-109624718 on Build GRCh38
rs778550799 Chr.X: 109624586-109624586 on Build GRCh38
rs768958435 Chr.X: 109624597-109624597 on Build GRCh38
rs41312935 Chr.X: 109624681-109624681 on Build GRCh38
rs72548776 Chr.X: 109624639-109624639 on Build GRCh38
rs767463268 Chr.X: 109624696-109624696 on Build GRCh38
rs757868800 Chr.X: 109624691-109624691 on Build GRCh38
rs1024077613 Chr.X: 109624565-109624565 on Build GRCh38
rs200723915 Chr.X: 109624745-109624745 on Build GRCh38
rs756286045 Chr.X: 109624704-109624704 on Build GRCh38
rs764993726 Chr.X: 109624675-109624675 on Build GRCh38
rs375008004 Chr.X: 109624716-109624716 on Build GRCh38
rs1018945439 Chr.X: 109624684-109624684 on Build GRCh38
rs749524117 Chr.X: 109624708-109624708 on Build GRCh38
rs746974037 Chr.X: 109624653-109624653 on Build GRCh38
rs372565167 Chr.X: 109624663-109624663 on Build GRCh38
rs72548778 Chr.X: 109624685-109624685 on Build GRCh38
rs758138171 Chr.X: 109624712-109624712 on Build GRCh38
rs762719236 Chr.X: 109624740-109624740 on Build GRCh38
rs374913815 Chr.X: 109624741-109624741 on Build GRCh38
rs72548777 Chr.X: 109624674-109624674 on Build GRCh38
rs773078028 Chr.X: 109624606-109624606 on Build GRCh38
rs891015394 Chr.X: 109624637-109624637 on Build GRCh38
rs763339959 Chr.X: 109624686-109624686 on Build GRCh38
rs764564753 Chr.X: 109624687-109624687 on Build GRCh38
rs775987548 Chr.X: 109624723-109624723 on Build GRCh38
rs951525284 Chr.X: 109624672-109624672 on Build GRCh38
rs369319019 Chr.X: 109624732-109624732 on Build GRCh38
rs982739595 Chr.X: 109624634-109624634 on Build GRCh38
rs761928628 Chr.X: 109624654-109624654 on Build GRCh38
rs760918742 Chr.X: 109624576-109624576 on Build GRCh38
rs748004704 Chr.X: 109624588-109624588 on Build GRCh38
rs770947110 Chr.X: 109624721-109624721 on Build GRCh38
rs759093005 Chr.X: 109624665-109624665 on Build GRCh38
rs757341707 Chr.X: 109624720-109624724 on Build GRCh38
rs969431597 Chr.X: 109624630-109624630 on Build GRCh38
rs752061790 Chr.X: 109624688-109624688 on Build GRCh38
rs1033061575 Chr.X: 109624713-109624713 on Build GRCh38
rs756398184 Chr.X: 109624577-109624577 on Build GRCh38
rs750371428 Chr.X: 109624736-109624736 on Build GRCh38

View Full Product Details