Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 11885585-11885852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAL
Entrez Gene ID
2774
Gene Name
G protein subunit alpha L
Gene Aliases
DYT25
UniGene
Hs.136295
Gene Chromosome Location
Chr.18: 11688956-11885685 on Build GRCh38
Transcripts
NM_001142339.2 NM_001261443.1 NM_001261444.1 NM_182978.3 NM_001242904.1 NM_001319154.1 NM_023075.5 NR_040241.1 NR_040242.3 NR_040243.1 XM_006722340.3 XM_006722341.3 XM_006722342.3 XM_006722343.3 XM_006722345.3 XM_006722346.2 XM_006722347.3 XM_006722348.2 XM_011525731.2 XM_011525732.1 XM_011525734.1 XM_017025920.1 XM_017025921.1 XM_017025922.1 XM_017025923.1 XM_017025924.1 XM_017025925.1 XM_017025926.1 XM_017025927.1 XM_017025928.1 XM_017025929.1 XM_017025930.1

Gene Symbol
MPPE1
Entrez Gene ID
65258
Gene Name
metallophosphoesterase 1
Gene Aliases
PGAP5
UniGene
Hs.712666
Gene Chromosome Location
Chr.18: 11883471-11908797 on Build GRCh38
Transcripts
NM_001142339.2 NM_001261443.1 NM_001261444.1 NM_182978.3 NM_001242904.1 NM_001319154.1 NM_023075.5 NR_040241.1 NR_040242.3 NR_040243.1 XM_006722340.3 XM_006722341.3 XM_006722342.3 XM_006722343.3 XM_006722345.3 XM_006722346.2 XM_006722347.3 XM_006722348.2 XM_011525731.2 XM_011525732.1 XM_011525734.1 XM_017025920.1 XM_017025921.1 XM_017025922.1 XM_017025923.1 XM_017025924.1 XM_017025925.1 XM_017025926.1 XM_017025927.1 XM_017025928.1 XM_017025929.1 XM_017025930.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6148257 c.962T>G p.V321G Chr.18: 11885722-11885722 on Build GRCh38
1387786 c.917G>A p.S306N Chr.18: 11885767-11885767 on Build GRCh38
4703080 c.888G>A p.P296P Chr.18: 11885796-11885796 on Build GRCh38
6871969 c.928G>A p.V310M Chr.18: 11885756-11885756 on Build GRCh38
5783582 c.934C>T p.H312Y Chr.18: 11885750-11885750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558957607 Chr.18: 11885766-11885766 on Build GRCh38
rs765333163 Chr.18: 11885761-11885761 on Build GRCh38
rs368111952 Chr.18: 11885645-11885645 on Build GRCh38
rs747813934 Chr.18: 11885744-11885744 on Build GRCh38
rs754090349 Chr.18: 11885747-11885748 on Build GRCh38
rs372251919 Chr.18: 11885795-11885795 on Build GRCh38
rs370831847 Chr.18: 11885760-11885760 on Build GRCh38
rs960997028 Chr.18: 11885734-11885734 on Build GRCh38
rs758601196 Chr.18: 11885723-11885723 on Build GRCh38
rs372742171 Chr.18: 11885703-11885703 on Build GRCh38
rs141361459 Chr.18: 11885747-11885747 on Build GRCh38
rs770164074 Chr.18: 11885821-11885821 on Build GRCh38
rs764267556 Chr.18: 11885743-11885743 on Build GRCh38
rs765303790 Chr.18: 11885710-11885710 on Build GRCh38
rs1039864091 Chr.18: 11885771-11885771 on Build GRCh38
rs926176134 Chr.18: 11885765-11885765 on Build GRCh38
rs749188944 Chr.18: 11885738-11885738 on Build GRCh38
rs761005446 Chr.18: 11885741-11885742 on Build GRCh38
rs767910786 Chr.18: 11885689-11885689 on Build GRCh38
rs780063778 Chr.18: 11885794-11885794 on Build GRCh38
rs776905221 Chr.18: 11885813-11885813 on Build GRCh38
rs775596031 Chr.18: 11885668-11885668 on Build GRCh38
rs546462592 Chr.18: 11885654-11885654 on Build GRCh38
rs375316149 Chr.18: 11885796-11885796 on Build GRCh38
rs752954066 Chr.18: 11885715-11885715 on Build GRCh38
rs1017014068 Chr.18: 11885808-11885808 on Build GRCh38
rs771582194 Chr.18: 11885804-11885804 on Build GRCh38
rs781195363 Chr.18: 11885640-11885640 on Build GRCh38
rs3208942 Chr.18: 11885682-11885682 on Build GRCh38
rs1060108 Chr.18: 11885781-11885781 on Build GRCh38
rs148506110 Chr.18: 11885833-11885833 on Build GRCh38
rs766824046 Chr.18: 11885699-11885699 on Build GRCh38
rs776091299 Chr.18: 11885828-11885828 on Build GRCh38
rs746729224 Chr.18: 11885806-11885806 on Build GRCh38
rs768062583 Chr.18: 11885671-11885671 on Build GRCh38
rs753083377 Chr.18: 11885642-11885642 on Build GRCh38
rs576465695 Chr.18: 11885740-11885740 on Build GRCh38
rs777338026 Chr.18: 11885805-11885805 on Build GRCh38
rs73945269 Chr.18: 11885773-11885773 on Build GRCh38
rs201590510 Chr.18: 11885782-11885782 on Build GRCh38
rs568274647 Chr.18: 11885695-11885695 on Build GRCh38
rs764276565 Chr.18: 11885831-11885831 on Build GRCh38
rs761060255 Chr.18: 11885698-11885698 on Build GRCh38
rs769880201 Chr.18: 11885759-11885759 on Build GRCh38
rs760053201 Chr.18: 11885819-11885819 on Build GRCh38
rs369058151 Chr.18: 11885829-11885829 on Build GRCh38
rs201131313 Chr.18: 11885724-11885724 on Build GRCh38
rs771269216 Chr.18: 11885812-11885812 on Build GRCh38
rs200161280 Chr.18: 11885733-11885733 on Build GRCh38
rs927058329 Chr.18: 11885701-11885701 on Build GRCh38
rs376272422 Chr.18: 11885756-11885756 on Build GRCh38
rs755998099 Chr.18: 11885732-11885732 on Build GRCh38
rs748780926 Chr.18: 11885670-11885670 on Build GRCh38
rs760970409 Chr.18: 11885748-11885748 on Build GRCh38
rs778346360 Chr.18: 11885741-11885741 on Build GRCh38
rs1005658649 Chr.18: 11885615-11885615 on Build GRCh38
rs536672004 Chr.18: 11885767-11885767 on Build GRCh38
rs745621767 Chr.18: 11885726-11885726 on Build GRCh38
rs755021078 Chr.18: 11885709-11885709 on Build GRCh38
rs773985657 Chr.18: 11885680-11885680 on Build GRCh38
rs559673104 Chr.18: 11885797-11885797 on Build GRCh38
rs16976814 Chr.18: 11885678-11885678 on Build GRCh38
rs747561914 Chr.18: 11885802-11885802 on Build GRCh38

View Full Product Details