Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42980945-42981204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RUNDC1
Entrez Gene ID
146923
Gene Name
RUN domain containing 1
Gene Aliases
-
UniGene
Hs.632255
Gene Chromosome Location
Chr.17: 42980510-42993692 on Build GRCh38
Transcripts
NM_001321381.1 NM_173079.3 XM_005257078.3 XM_005257080.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4650416 c.490G>T p.E164* Chr.17: 42981066-42981066 on Build GRCh38
7256966 c.432C>A p.G144G Chr.17: 42981008-42981008 on Build GRCh38
3755558 c.478T>C p.W160R Chr.17: 42981054-42981054 on Build GRCh38
1734434 c.389C>A p.A130D Chr.17: 42980965-42980965 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762389615 Chr.17: 42981087-42981087 on Build GRCh38
rs1010650507 Chr.17: 42980997-42980997 on Build GRCh38
rs999570905 Chr.17: 42981044-42981044 on Build GRCh38
rs747204117 Chr.17: 42981055-42981055 on Build GRCh38
rs751226206 Chr.17: 42981004-42981004 on Build GRCh38
rs559051216 Chr.17: 42981095-42981095 on Build GRCh38
rs765265688 Chr.17: 42980970-42980970 on Build GRCh38
rs760923216 Chr.17: 42981071-42981071 on Build GRCh38
rs939251203 Chr.17: 42980963-42980963 on Build GRCh38
rs767250445 Chr.17: 42981159-42981159 on Build GRCh38
rs966892293 Chr.17: 42981180-42981180 on Build GRCh38
rs537037839 Chr.17: 42981077-42981077 on Build GRCh38
rs776264058 Chr.17: 42981063-42981063 on Build GRCh38
rs767630191 Chr.17: 42980984-42980985 on Build GRCh38
rs772087875 Chr.17: 42981065-42981065 on Build GRCh38
rs759373874 Chr.17: 42980964-42980964 on Build GRCh38
rs781273982 Chr.17: 42981056-42981056 on Build GRCh38
rs779605527 Chr.17: 42981045-42981045 on Build GRCh38
rs745990273 Chr.17: 42981127-42981127 on Build GRCh38
rs755420359 Chr.17: 42981043-42981043 on Build GRCh38
rs771239383 Chr.17: 42981061-42981061 on Build GRCh38
rs752759292 Chr.17: 42981033-42981042 on Build GRCh38
rs978283166 Chr.17: 42981184-42981184 on Build GRCh38
rs756432986 Chr.17: 42981005-42981005 on Build GRCh38
rs552222 Chr.17: 42980988-42980988 on Build GRCh38
rs952253597 Chr.17: 42981118-42981118 on Build GRCh38
rs775426856 Chr.17: 42981086-42981086 on Build GRCh38
rs754121834 Chr.17: 42981175-42981175 on Build GRCh38
rs902166519 Chr.17: 42981029-42981029 on Build GRCh38
rs989537306 Chr.17: 42981169-42981169 on Build GRCh38
rs766648925 Chr.17: 42981009-42981009 on Build GRCh38
rs1708875 Chr.17: 42981054-42981054 on Build GRCh38
rs915316728 Chr.17: 42981172-42981172 on Build GRCh38
rs748202071 Chr.17: 42981048-42981048 on Build GRCh38
rs763598952 Chr.17: 42981091-42981091 on Build GRCh38
rs996393883 Chr.17: 42981092-42981092 on Build GRCh38
rs769575618 Chr.17: 42981080-42981080 on Build GRCh38
rs754286789 Chr.17: 42981025-42981025 on Build GRCh38
rs774946106 Chr.17: 42980987-42980987 on Build GRCh38

View Full Product Details