Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 32114463-32114732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPAST
Entrez Gene ID
6683
Gene Name
spastin
Gene Aliases
ADPSP, FSP2, SPG4
UniGene
Hs.468091
Gene Chromosome Location
Chr.2: 32063551-32157637 on Build GRCh38
Transcripts
NM_014946.3 NM_199436.1 XM_005264516.4 XM_011533067.2 XM_017004777.1 XM_017004778.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4838067 c.726T>C p.T242T Chr.2: 32114681-32114681 on Build GRCh38
4505619 c.697C>T p.P233S Chr.2: 32114652-32114652 on Build GRCh38
6477844 c.759G>A p.M253I Chr.2: 32114714-32114714 on Build GRCh38
6788038 c.758T>G p.M253R Chr.2: 32114713-32114713 on Build GRCh38
6157977 c.689G>C p.G230A Chr.2: 32114644-32114644 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140094231 Chr.2: 32114667-32114667 on Build GRCh38
rs561910912 Chr.2: 32114569-32114569 on Build GRCh38
rs577586915 Chr.2: 32114509-32114509 on Build GRCh38
rs1005545731 Chr.2: 32114593-32114593 on Build GRCh38
rs751612051 Chr.2: 32114666-32114666 on Build GRCh38
rs902256615 Chr.2: 32114518-32114518 on Build GRCh38
rs766843588 Chr.2: 32114641-32114641 on Build GRCh38
rs753598913 Chr.2: 32114691-32114691 on Build GRCh38
rs770841090 Chr.2: 32114594-32114594 on Build GRCh38
rs188446799 Chr.2: 32114602-32114602 on Build GRCh38
rs540249161 Chr.2: 32114524-32114524 on Build GRCh38
rs756026679 Chr.2: 32114680-32114680 on Build GRCh38
rs201395991 Chr.2: 32114698-32114698 on Build GRCh38
rs374954461 Chr.2: 32114597-32114597 on Build GRCh38
rs762209579 Chr.2: 32114613-32114613 on Build GRCh38
rs939863288 Chr.2: 32114522-32114522 on Build GRCh38
rs896334403 Chr.2: 32114557-32114557 on Build GRCh38
rs939245156 Chr.2: 32114539-32114539 on Build GRCh38
rs757176714 Chr.2: 32114697-32114697 on Build GRCh38
rs752959206 Chr.2: 32114673-32114673 on Build GRCh38
rs955517877 Chr.2: 32114683-32114683 on Build GRCh38
rs765581987 Chr.2: 32114614-32114614 on Build GRCh38
rs550646462 Chr.2: 32114617-32114617 on Build GRCh38
rs202209866 Chr.2: 32114629-32114629 on Build GRCh38
rs1055442009 Chr.2: 32114563-32114563 on Build GRCh38
rs946390341 Chr.2: 32114486-32114486 on Build GRCh38
rs573069278 Chr.2: 32114534-32114534 on Build GRCh38
rs745422831 Chr.2: 32114709-32114709 on Build GRCh38
rs775105475 Chr.2: 32114605-32114605 on Build GRCh38
rs11691939 Chr.2: 32114492-32114492 on Build GRCh38
rs895173062 Chr.2: 32114576-32114576 on Build GRCh38
rs749163368 Chr.2: 32114587-32114587 on Build GRCh38
rs560319865 Chr.2: 32114530-32114530 on Build GRCh38
rs550256815 Chr.2: 32114595-32114595 on Build GRCh38
rs541803680 Chr.2: 32114548-32114548 on Build GRCh38
rs1036783300 Chr.2: 32114529-32114529 on Build GRCh38
rs767891833 Chr.2: 32114668-32114668 on Build GRCh38

View Full Product Details