Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 33503370-33503603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTI2
Entrez Gene ID
80185
Gene Name
TELO2 interacting protein 2
Gene Aliases
C8orf41, MRT39
UniGene
Hs.742048
Gene Chromosome Location
Chr.8: 33498509-33513601 on Build GRCh38
Transcripts
NM_001102401.2 NM_001265581.1 NM_025115.3 XM_011544665.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6271016 c.1243C>A p.Q415K Chr.8: 33503445-33503445 on Build GRCh38
6811377 c.1211T>C p.L404P Chr.8: 33503477-33503477 on Build GRCh38
5459366 c.1132G>A p.V378I Chr.8: 33503556-33503556 on Build GRCh38
3834610 c.1225A>C p.T409P Chr.8: 33503463-33503463 on Build GRCh38
454487 c.1154A>T p.E385V Chr.8: 33503534-33503534 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891841113 Chr.8: 33503504-33503504 on Build GRCh38
rs771416460 Chr.8: 33503503-33503503 on Build GRCh38
rs866181892 Chr.8: 33503543-33503543 on Build GRCh38
rs971346120 Chr.8: 33503552-33503552 on Build GRCh38
rs780879074 Chr.8: 33503468-33503468 on Build GRCh38
rs142647692 Chr.8: 33503471-33503471 on Build GRCh38
rs564262431 Chr.8: 33503572-33503572 on Build GRCh38
rs764888270 Chr.8: 33503409-33503409 on Build GRCh38
rs967544161 Chr.8: 33503444-33503444 on Build GRCh38
rs1009406044 Chr.8: 33503529-33503530 on Build GRCh38
rs766391051 Chr.8: 33503561-33503561 on Build GRCh38
rs148492377 Chr.8: 33503469-33503469 on Build GRCh38
rs150984360 Chr.8: 33503498-33503498 on Build GRCh38
rs200629022 Chr.8: 33503528-33503528 on Build GRCh38
rs1009167526 Chr.8: 33503431-33503431 on Build GRCh38
rs775803386 Chr.8: 33503527-33503527 on Build GRCh38
rs751082403 Chr.8: 33503562-33503562 on Build GRCh38
rs778746953 Chr.8: 33503571-33503571 on Build GRCh38
rs759628656 Chr.8: 33503515-33503515 on Build GRCh38
rs367640273 Chr.8: 33503408-33503408 on Build GRCh38
rs750413886 Chr.8: 33503412-33503412 on Build GRCh38
rs778045782 Chr.8: 33503495-33503495 on Build GRCh38
rs917260096 Chr.8: 33503526-33503526 on Build GRCh38
rs556828933 Chr.8: 33503529-33503529 on Build GRCh38
rs774479699 Chr.8: 33503509-33503509 on Build GRCh38
rs772232196 Chr.8: 33503522-33503522 on Build GRCh38
rs751113227 Chr.8: 33503441-33503441 on Build GRCh38
rs754543529 Chr.8: 33503567-33503567 on Build GRCh38
rs985999682 Chr.8: 33503563-33503563 on Build GRCh38
rs1033589421 Chr.8: 33503491-33503491 on Build GRCh38
rs758341057 Chr.8: 33503418-33503418 on Build GRCh38
rs753615989 Chr.8: 33503401-33503401 on Build GRCh38
rs140977711 Chr.8: 33503553-33503553 on Build GRCh38
rs754630288 Chr.8: 33503448-33503448 on Build GRCh38
rs199664580 Chr.8: 33503438-33503438 on Build GRCh38
rs767773664 Chr.8: 33503396-33503396 on Build GRCh38
rs765124924 Chr.8: 33503550-33503550 on Build GRCh38
rs200737331 Chr.8: 33503531-33503531 on Build GRCh38

View Full Product Details