Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 52157639-52157886 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA10
Entrez Gene ID
56934
Gene Name
carbonic anhydrase 10
Gene Aliases
CA-RPX, CARPX, HUCEP-15
UniGene
Hs.463466
Gene Chromosome Location
Chr.17: 51630313-52160017 on Build GRCh38
Transcripts
NM_001082533.1 NM_001082534.1 NM_020178.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
334064 c.14G>A p.W5* Chr.17: 52157773-52157773 on Build GRCh38
383192 c.31C>A p.L11I Chr.17: 52157756-52157756 on Build GRCh38
6464995 c.19G>A p.V7M Chr.17: 52157768-52157768 on Build GRCh38
344554 c.45C>T p.F15F Chr.17: 52157742-52157742 on Build GRCh38
5852997 c.15G>A p.W5* Chr.17: 52157772-52157772 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs908815977 Chr.17: 52157760-52157760 on Build GRCh38
rs28571897 Chr.17: 52157711-52157711 on Build GRCh38
rs775003760 Chr.17: 52157768-52157768 on Build GRCh38
rs11655970 Chr.17: 52157782-52157782 on Build GRCh38
rs767601274 Chr.17: 52157729-52157729 on Build GRCh38
rs201312360 Chr.17: 52157704-52157704 on Build GRCh38
rs760664193 Chr.17: 52157836-52157836 on Build GRCh38
rs752658560 Chr.17: 52157730-52157730 on Build GRCh38
rs777617655 Chr.17: 52157828-52157828 on Build GRCh38
rs770646683 Chr.17: 52157816-52157816 on Build GRCh38
rs768978296 Chr.17: 52157796-52157796 on Build GRCh38
rs371529572 Chr.17: 52157722-52157722 on Build GRCh38
rs201187222 Chr.17: 52157717-52157717 on Build GRCh38
rs1013872136 Chr.17: 52157669-52157669 on Build GRCh38
rs895592776 Chr.17: 52157664-52157664 on Build GRCh38
rs765890082 Chr.17: 52157720-52157720 on Build GRCh38
rs759452367 Chr.17: 52157828-52157828 on Build GRCh38
rs367996538 Chr.17: 52157714-52157714 on Build GRCh38
rs753789532 Chr.17: 52157744-52157744 on Build GRCh38
rs774115154 Chr.17: 52157819-52157819 on Build GRCh38
rs746273689 Chr.17: 52157765-52157765 on Build GRCh38
rs756317806 Chr.17: 52157738-52157738 on Build GRCh38
rs954816270 Chr.17: 52157808-52157808 on Build GRCh38
rs778967703 Chr.17: 52157748-52157748 on Build GRCh38
rs372020862 Chr.17: 52157701-52157701 on Build GRCh38
rs775630788 Chr.17: 52157832-52157832 on Build GRCh38
rs140850689 Chr.17: 52157741-52157741 on Build GRCh38
rs772792597 Chr.17: 52157798-52157798 on Build GRCh38
rs537580773 Chr.17: 52157827-52157828 on Build GRCh38
rs749047298 Chr.17: 52157836-52157836 on Build GRCh38
rs767403123 Chr.17: 52157830-52157830 on Build GRCh38
rs368674548 Chr.17: 52157687-52157687 on Build GRCh38
rs751392227 Chr.17: 52157721-52157721 on Build GRCh38
rs757526132 Chr.17: 52157746-52157746 on Build GRCh38
rs762671330 Chr.17: 52157715-52157715 on Build GRCh38
rs748696349 Chr.17: 52157806-52157806 on Build GRCh38
rs62070291 Chr.17: 52157844-52157844 on Build GRCh38
rs34449660 Chr.17: 52157721-52157722 on Build GRCh38
rs763876734 Chr.17: 52157837-52157837 on Build GRCh38
rs748246451 Chr.17: 52157698-52157698 on Build GRCh38

View Full Product Details