Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 23958174-23958409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBXN2A
Entrez Gene ID
165324
Gene Name
UBX domain protein 2A
Gene Aliases
UBXD4
UniGene
Hs.591576
Gene Chromosome Location
Chr.2: 23927201-24001000 on Build GRCh38
Transcripts
NM_181713.3 XM_005264166.4 XM_005264167.4 XM_005264168.4 XM_005264169.4 XM_011532633.2 XM_017003517.1 XM_017003518.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5679640 c.10G>T p.V4L Chr.2: 23958324-23958324 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767955819 Chr.2: 23958326-23958326 on Build GRCh38
rs763663893 Chr.2: 23958270-23958270 on Build GRCh38
rs1028202648 Chr.2: 23958229-23958229 on Build GRCh38
rs12616678 Chr.2: 23958289-23958289 on Build GRCh38
rs759705143 Chr.2: 23958324-23958324 on Build GRCh38
rs1031878841 Chr.2: 23958298-23958298 on Build GRCh38
rs955391684 Chr.2: 23958260-23958260 on Build GRCh38
rs778699327 Chr.2: 23958285-23958285 on Build GRCh38
rs1034844528 Chr.2: 23958342-23958342 on Build GRCh38
rs748502540 Chr.2: 23958232-23958232 on Build GRCh38
rs762782885 Chr.2: 23958275-23958289 on Build GRCh38
rs753338550 Chr.2: 23958271-23958271 on Build GRCh38
rs761372466 Chr.2: 23958376-23958376 on Build GRCh38
rs763655293 Chr.2: 23958310-23958311 on Build GRCh38
rs776799658 Chr.2: 23958274-23958277 on Build GRCh38
rs994294829 Chr.2: 23958271-23958271 on Build GRCh38
rs757025298 Chr.2: 23958278-23958278 on Build GRCh38
rs980766085 Chr.2: 23958333-23958333 on Build GRCh38
rs766111539 Chr.2: 23958292-23958293 on Build GRCh38
rs560402186 Chr.2: 23958226-23958226 on Build GRCh38
rs370870768 Chr.2: 23958307-23958307 on Build GRCh38
rs150887344 Chr.2: 23958337-23958337 on Build GRCh38
rs758239517 Chr.2: 23958288-23958288 on Build GRCh38
rs761257103 Chr.2: 23958354-23958354 on Build GRCh38
rs901231898 Chr.2: 23958235-23958235 on Build GRCh38
rs1041255219 Chr.2: 23958208-23958208 on Build GRCh38
rs768422579 Chr.2: 23958295-23958295 on Build GRCh38
rs200322972 Chr.2: 23958323-23958323 on Build GRCh38
rs745694974 Chr.2: 23958286-23958286 on Build GRCh38
rs746972815 Chr.2: 23958291-23958291 on Build GRCh38
rs542792859 Chr.2: 23958306-23958306 on Build GRCh38
rs753463289 Chr.2: 23958357-23958357 on Build GRCh38
rs113328530 Chr.2: 23958305-23958305 on Build GRCh38
rs764492312 Chr.2: 23958355-23958355 on Build GRCh38
rs987948396 Chr.2: 23958360-23958360 on Build GRCh38

View Full Product Details