Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 185922426-185922653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMCN1
Entrez Gene ID
83872
Gene Name
hemicentin 1
Gene Aliases
ARMD1, FBLN6, FIBL-6, FIBL6
UniGene
Hs.58877
Gene Chromosome Location
Chr.1: 185734526-186190953 on Build GRCh38
Transcripts
NM_031935.2 XM_011510038.2 XM_011510041.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1172076 c.967T>C p.F323L Chr.1: 185922445-185922445 on Build GRCh38
254724 c.958C>T p.R320* Chr.1: 185922436-185922436 on Build GRCh38
4514429 c.973C>T p.R325* Chr.1: 185922451-185922451 on Build GRCh38
5010169 c.951T>C p.I317I Chr.1: 185922429-185922429 on Build GRCh38
4612990 c.993_994insA p.T334fs*25 Chr.1: 185922471-185922472 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370021314 Chr.1: 185922451-185922451 on Build GRCh38
rs538430915 Chr.1: 185922582-185922582 on Build GRCh38
rs958824151 Chr.1: 185922574-185922574 on Build GRCh38
rs769155073 Chr.1: 185922490-185922490 on Build GRCh38
rs868658018 Chr.1: 185922479-185922482 on Build GRCh38
rs751043199 Chr.1: 185922461-185922461 on Build GRCh38
rs749747442 Chr.1: 185922511-185922511 on Build GRCh38
rs779005077 Chr.1: 185922513-185922513 on Build GRCh38
rs368610007 Chr.1: 185922539-185922539 on Build GRCh38
rs745928751 Chr.1: 185922492-185922493 on Build GRCh38
rs917441441 Chr.1: 185922609-185922609 on Build GRCh38
rs202206115 Chr.1: 185922452-185922452 on Build GRCh38
rs756883609 Chr.1: 185922462-185922462 on Build GRCh38
rs186852890 Chr.1: 185922567-185922567 on Build GRCh38
rs780737748 Chr.1: 185922466-185922466 on Build GRCh38
rs748211677 Chr.1: 185922514-185922514 on Build GRCh38
rs199547255 Chr.1: 185922507-185922507 on Build GRCh38
rs772237229 Chr.1: 185922522-185922522 on Build GRCh38
rs1013243738 Chr.1: 185922594-185922594 on Build GRCh38
rs769058338 Chr.1: 185922512-185922512 on Build GRCh38
rs750140043 Chr.1: 185922470-185922470 on Build GRCh38
rs138572318 Chr.1: 185922482-185922482 on Build GRCh38

View Full Product Details