Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 93797524-93797781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MED17
Entrez Gene ID
9440
Gene Name
mediator complex subunit 17
Gene Aliases
CRSP6, CRSP77, DRIP80, TRAP80
UniGene
Hs.444931
Gene Chromosome Location
Chr.11: 93784239-93813330 on Build GRCh38
Transcripts
NM_004268.4 XM_011543068.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5821506 c.1241A>T p.D414V Chr.11: 93797632-93797632 on Build GRCh38
7150271 c.1277G>T p.G426V Chr.11: 93797668-93797668 on Build GRCh38
6710858 c.1162A>G p.S388G Chr.11: 93797553-93797553 on Build GRCh38
4037664 c.1172T>C p.M391T Chr.11: 93797563-93797563 on Build GRCh38
7424353 c.1215G>T p.M405I Chr.11: 93797606-93797606 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs36120755 Chr.11: 93797651-93797651 on Build GRCh38
rs746638316 Chr.11: 93797591-93797591 on Build GRCh38
rs748031133 Chr.11: 93797571-93797571 on Build GRCh38
rs1055449249 Chr.11: 93797655-93797655 on Build GRCh38
rs778974560 Chr.11: 93797711-93797711 on Build GRCh38
rs376870817 Chr.11: 93797607-93797607 on Build GRCh38
rs571873238 Chr.11: 93797709-93797709 on Build GRCh38
rs939603700 Chr.11: 93797619-93797619 on Build GRCh38
rs200531476 Chr.11: 93797746-93797746 on Build GRCh38
rs752544714 Chr.11: 93797727-93797727 on Build GRCh38
rs767747153 Chr.11: 93797658-93797658 on Build GRCh38
rs200391569 Chr.11: 93797735-93797735 on Build GRCh38
rs566018883 Chr.11: 93797581-93797581 on Build GRCh38
rs534953424 Chr.11: 93797600-93797600 on Build GRCh38
rs777520439 Chr.11: 93797583-93797583 on Build GRCh38
rs966671158 Chr.11: 93797608-93797608 on Build GRCh38
rs140719424 Chr.11: 93797614-93797614 on Build GRCh38
rs770708734 Chr.11: 93797747-93797747 on Build GRCh38
rs915231908 Chr.11: 93797606-93797606 on Build GRCh38
rs916785618 Chr.11: 93797661-93797661 on Build GRCh38
rs769222927 Chr.11: 93797615-93797615 on Build GRCh38
rs766549983 Chr.11: 93797696-93797696 on Build GRCh38
rs16919387 Chr.11: 93797731-93797731 on Build GRCh38
rs930384502 Chr.11: 93797691-93797691 on Build GRCh38
rs887554981 Chr.11: 93797698-93797698 on Build GRCh38
rs770655353 Chr.11: 93797599-93797599 on Build GRCh38
rs763249105 Chr.11: 93797688-93797691 on Build GRCh38
rs778651154 Chr.11: 93797558-93797558 on Build GRCh38
rs866797955 Chr.11: 93797595-93797595 on Build GRCh38
rs750692497 Chr.11: 93797660-93797660 on Build GRCh38
rs777029450 Chr.11: 93797742-93797742 on Build GRCh38
rs760909439 Chr.11: 93797689-93797689 on Build GRCh38
rs753921272 Chr.11: 93797703-93797703 on Build GRCh38
rs774709960 Chr.11: 93797639-93797639 on Build GRCh38
rs773492828 Chr.11: 93797599-93797600 on Build GRCh38

View Full Product Details