Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 43033728-43033959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RIT2
Entrez Gene ID
6014
Gene Name
Ras like without CAAX 2
Gene Aliases
RIBA, RIN, ROC2
UniGene
Hs.464985
Gene Chromosome Location
Chr.18: 42743218-43115692 on Build GRCh38
Transcripts
NM_001272077.1 NM_002930.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7259914 c.104-8_104-7insT p.? Chr.18: 43033874-43033875 on Build GRCh38
3525802 c.136C>T p.P46S Chr.18: 43033835-43033835 on Build GRCh38
3742620 c.113T>G p.M38R Chr.18: 43033858-43033858 on Build GRCh38
1663930 c.104C>A p.A35E Chr.18: 43033867-43033867 on Build GRCh38
4417738 c.104-8T>C p.? Chr.18: 43033875-43033875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376527258 Chr.18: 43033768-43033768 on Build GRCh38
rs776728832 Chr.18: 43033851-43033851 on Build GRCh38
rs112790285 Chr.18: 43033874-43033875 on Build GRCh38
rs754909307 Chr.18: 43033913-43033913 on Build GRCh38
rs903382036 Chr.18: 43033889-43033889 on Build GRCh38
rs750145350 Chr.18: 43033891-43033891 on Build GRCh38
rs771120789 Chr.18: 43033844-43033844 on Build GRCh38
rs369613443 Chr.18: 43033771-43033771 on Build GRCh38
rs190080771 Chr.18: 43033917-43033917 on Build GRCh38
rs376172944 Chr.18: 43033897-43033897 on Build GRCh38
rs755270302 Chr.18: 43033875-43033875 on Build GRCh38
rs753659593 Chr.18: 43033898-43033898 on Build GRCh38
rs551330973 Chr.18: 43033871-43033871 on Build GRCh38
rs759963635 Chr.18: 43033855-43033855 on Build GRCh38
rs747093603 Chr.18: 43033818-43033818 on Build GRCh38
rs755872002 Chr.18: 43033892-43033892 on Build GRCh38
rs201571045 Chr.18: 43033878-43033878 on Build GRCh38
rs112340206 Chr.18: 43033926-43033926 on Build GRCh38
rs778722775 Chr.18: 43033916-43033916 on Build GRCh38
rs12954451 Chr.18: 43033875-43033875 on Build GRCh38
rs770344980 Chr.18: 43033856-43033856 on Build GRCh38
rs563155930 Chr.18: 43033775-43033775 on Build GRCh38
rs766505085 Chr.18: 43033873-43033874 on Build GRCh38
rs186095570 Chr.18: 43033890-43033890 on Build GRCh38
rs143764572 Chr.18: 43033863-43033863 on Build GRCh38
rs779963531 Chr.18: 43033778-43033778 on Build GRCh38
rs761231300 Chr.18: 43033882-43033882 on Build GRCh38
rs772117492 Chr.18: 43033788-43033788 on Build GRCh38
rs373158597 Chr.18: 43033787-43033787 on Build GRCh38
rs751038765 Chr.18: 43033877-43033877 on Build GRCh38
rs777863748 Chr.18: 43033814-43033814 on Build GRCh38
rs755963618 Chr.18: 43033776-43033776 on Build GRCh38

View Full Product Details