Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 52136491-52136764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST18
Entrez Gene ID
9705
Gene Name
ST18, C2H2C-type zinc finger
Gene Aliases
NZF3, ZC2H2C3, ZC2HC10, ZNF387
UniGene
Hs.655499
Gene Chromosome Location
Chr.8: 52110832-52409879 on Build GRCh38
Transcripts
NM_014682.2 XM_006716487.1 XM_011517629.1 XM_011517631.1 XM_011517632.1 XM_011517633.1 XM_011517634.1 XM_011517635.1 XM_011517636.2 XM_011517637.1 XM_011517638.2 XM_011517641.1 XM_011517642.1 XM_017014047.1 XM_017014048.1 XM_017014049.1 XM_017014050.1 XM_017014051.1 XM_017014052.1 XM_017014053.1 XM_017014054.1 XM_017014055.1 XM_017014056.1 XM_017014057.1 XM_017014058.1 XM_017014059.1 XM_017014060.1 XM_017014061.1 XM_017014062.1 XM_017014063.1 XM_017014064.1 XM_017014065.1 XM_017014066.1 XM_017014067.1 XM_017014068.1 XM_017014069.1 XM_017014070.1 XM_017014071.1 XM_017014072.1 XM_017014073.1 XM_017014074.1 XM_017014075.1 XM_017014076.1 XM_017014077.1 XM_017014078.1 XM_017014079.1 XM_017014080.1 XM_017014081.1 XM_017014082.1 XM_017014083.1 XM_017014084.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1100443 c.2237C>A p.S746Y Chr.8: 52136653-52136653 on Build GRCh38
3649636 c.2240G>A p.G747E Chr.8: 52136650-52136650 on Build GRCh38
1330957 c.2287T>A p.S763T Chr.8: 52136603-52136603 on Build GRCh38
1100442 c.2291A>G p.Q764R Chr.8: 52136599-52136599 on Build GRCh38
751238 c.2270C>T p.S757F Chr.8: 52136620-52136620 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199527592 Chr.8: 52136598-52136598 on Build GRCh38
rs1011077202 Chr.8: 52136581-52136581 on Build GRCh38
rs377748068 Chr.8: 52136697-52136697 on Build GRCh38
rs777430747 Chr.8: 52136687-52136687 on Build GRCh38
rs566802977 Chr.8: 52136520-52136520 on Build GRCh38
rs764165724 Chr.8: 52136590-52136590 on Build GRCh38
rs376583231 Chr.8: 52136582-52136582 on Build GRCh38
rs1003233337 Chr.8: 52136682-52136682 on Build GRCh38
rs768392472 Chr.8: 52136674-52136675 on Build GRCh38
rs769430029 Chr.8: 52136568-52136568 on Build GRCh38
rs774149113 Chr.8: 52136679-52136680 on Build GRCh38
rs770925737 Chr.8: 52136579-52136579 on Build GRCh38
rs754469462 Chr.8: 52136664-52136664 on Build GRCh38
rs369779894 Chr.8: 52136602-52136602 on Build GRCh38
rs780931336 Chr.8: 52136555-52136555 on Build GRCh38
rs375733219 Chr.8: 52136548-52136548 on Build GRCh38
rs191343249 Chr.8: 52136733-52136733 on Build GRCh38
rs894015340 Chr.8: 52136575-52136575 on Build GRCh38
rs746911526 Chr.8: 52136634-52136634 on Build GRCh38
rs77869269 Chr.8: 52136702-52136702 on Build GRCh38
rs762627588 Chr.8: 52136675-52136675 on Build GRCh38
rs533967271 Chr.8: 52136523-52136523 on Build GRCh38
rs780782887 Chr.8: 52136672-52136672 on Build GRCh38
rs758968851 Chr.8: 52136625-52136625 on Build GRCh38
rs559716321 Chr.8: 52136692-52136692 on Build GRCh38
rs780050064 Chr.8: 52136589-52136589 on Build GRCh38
rs770727464 Chr.8: 52136699-52136699 on Build GRCh38
rs959065824 Chr.8: 52136732-52136732 on Build GRCh38
rs970835127 Chr.8: 52136519-52136519 on Build GRCh38
rs747838308 Chr.8: 52136567-52136567 on Build GRCh38
rs752272849 Chr.8: 52136681-52136681 on Build GRCh38
rs560471851 Chr.8: 52136698-52136698 on Build GRCh38
rs779679577 Chr.8: 52136547-52136547 on Build GRCh38
rs558519508 Chr.8: 52136529-52136529 on Build GRCh38
rs902284071 Chr.8: 52136671-52136671 on Build GRCh38
rs372876218 Chr.8: 52136534-52136534 on Build GRCh38
rs369100332 Chr.8: 52136549-52136549 on Build GRCh38
rs762115404 Chr.8: 52136601-52136601 on Build GRCh38
rs538341938 Chr.8: 52136540-52136540 on Build GRCh38
rs750839909 Chr.8: 52136604-52136604 on Build GRCh38
rs773110461 Chr.8: 52136569-52136569 on Build GRCh38
rs752121149 Chr.8: 52136662-52136662 on Build GRCh38
rs767047350 Chr.8: 52136658-52136658 on Build GRCh38
rs868740767 Chr.8: 52136619-52136619 on Build GRCh38
rs929349706 Chr.8: 52136524-52136524 on Build GRCh38
rs750844542 Chr.8: 52136539-52136539 on Build GRCh38

View Full Product Details