Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 106628785-106629034 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RTN4IP1
Entrez Gene ID
84816
Gene Name
reticulon 4 interacting protein 1
Gene Aliases
NIMP, OPA10
UniGene
Hs.155839
Gene Chromosome Location
Chr.6: 106559237-106630921 on Build GRCh38
Transcripts
NM_001318746.1 NM_032730.5 XM_011536192.2 XM_017011375.1 XM_017011376.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6770423 c.172C>T p.R58* Chr.6: 106628850-106628850 on Build GRCh38
164121 c.62G>A p.R21K Chr.6: 106628960-106628960 on Build GRCh38
5737717 c.89T>C p.V30A Chr.6: 106628933-106628933 on Build GRCh38
1697732 c.82C>T p.P28S Chr.6: 106628940-106628940 on Build GRCh38
4818450 c.172C>A p.R58R Chr.6: 106628850-106628850 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755748311 Chr.6: 106628905-106628905 on Build GRCh38
rs1023103707 Chr.6: 106628993-106628993 on Build GRCh38
rs759983578 Chr.6: 106628975-106628975 on Build GRCh38
rs753704233 Chr.6: 106628908-106628915 on Build GRCh38
rs150345358 Chr.6: 106628892-106628892 on Build GRCh38
rs753418145 Chr.6: 106628999-106628999 on Build GRCh38
rs1031608505 Chr.6: 106628991-106628991 on Build GRCh38
rs146158799 Chr.6: 106628983-106628983 on Build GRCh38
rs142241460 Chr.6: 106628947-106628947 on Build GRCh38
rs770620459 Chr.6: 106628915-106628915 on Build GRCh38
rs758659907 Chr.6: 106628869-106628869 on Build GRCh38
rs781777452 Chr.6: 106628895-106628895 on Build GRCh38
rs887117780 Chr.6: 106628913-106628913 on Build GRCh38
rs765442352 Chr.6: 106628835-106628835 on Build GRCh38
rs773976896 Chr.6: 106628918-106628918 on Build GRCh38
rs747627789 Chr.6: 106628903-106628903 on Build GRCh38
rs756809073 Chr.6: 106629001-106629001 on Build GRCh38
rs369636486 Chr.6: 106628958-106628958 on Build GRCh38
rs771974139 Chr.6: 106628945-106628945 on Build GRCh38
rs748829386 Chr.6: 106628910-106628910 on Build GRCh38
rs934310132 Chr.6: 106628816-106628816 on Build GRCh38
rs775475545 Chr.6: 106628946-106628946 on Build GRCh38
rs754753433 Chr.6: 106628917-106628917 on Build GRCh38
rs573671423 Chr.6: 106628974-106628974 on Build GRCh38
rs777352115 Chr.6: 106628998-106628998 on Build GRCh38
rs766649336 Chr.6: 106628966-106628966 on Build GRCh38
rs994688101 Chr.6: 106628902-106628902 on Build GRCh38
rs888483093 Chr.6: 106628909-106628909 on Build GRCh38
rs371469772 Chr.6: 106629006-106629006 on Build GRCh38
rs768131106 Chr.6: 106628982-106628982 on Build GRCh38
rs766589627 Chr.6: 106628874-106628874 on Build GRCh38
rs201266101 Chr.6: 106628956-106628956 on Build GRCh38
rs939243610 Chr.6: 106628870-106628870 on Build GRCh38
rs769807848 Chr.6: 106628950-106628950 on Build GRCh38
rs369908058 Chr.6: 106628828-106628828 on Build GRCh38
rs1036171818 Chr.6: 106628878-106628878 on Build GRCh38
rs750702643 Chr.6: 106628837-106628837 on Build GRCh38
rs1044673675 Chr.6: 106628815-106628815 on Build GRCh38
rs943206705 Chr.6: 106628836-106628836 on Build GRCh38
rs766393425 Chr.6: 106628903-106628904 on Build GRCh38
rs1987623 Chr.6: 106628908-106628908 on Build GRCh38
rs763048901 Chr.6: 106628963-106628963 on Build GRCh38
rs768561517 Chr.6: 106628914-106628914 on Build GRCh38
rs776642156 Chr.6: 106628823-106628823 on Build GRCh38
rs745696702 Chr.6: 106628939-106628939 on Build GRCh38
rs867884074 Chr.6: 106628981-106628981 on Build GRCh38
rs751989550 Chr.6: 106628884-106628884 on Build GRCh38
rs1824619 Chr.6: 106628821-106628821 on Build GRCh38

View Full Product Details