Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32310184-32310443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF3B
Entrez Gene ID
9371
Gene Name
kinesin family member 3B
Gene Aliases
FLA8, HH0048, KLP-11
UniGene
Hs.369670
Gene Chromosome Location
Chr.20: 32277640-32335011 on Build GRCh38
Transcripts
NM_004798.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1025703 c.460G>T p.D154Y Chr.20: 32310237-32310237 on Build GRCh38
3725115 c.618C>G p.T206T Chr.20: 32310395-32310395 on Build GRCh38
1025704 c.587T>C p.V196A Chr.20: 32310364-32310364 on Build GRCh38
227393 c.589G>A p.G197R Chr.20: 32310366-32310366 on Build GRCh38
1025705 c.601C>T p.R201C Chr.20: 32310378-32310378 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754158662 Chr.20: 32310269-32310269 on Build GRCh38
rs762187050 Chr.20: 32310377-32310377 on Build GRCh38
rs774694208 Chr.20: 32310205-32310205 on Build GRCh38
rs753966350 Chr.20: 32310395-32310395 on Build GRCh38
rs1033380046 Chr.20: 32310410-32310410 on Build GRCh38
rs772439368 Chr.20: 32310378-32310378 on Build GRCh38
rs745587157 Chr.20: 32310326-32310326 on Build GRCh38
rs28364805 Chr.20: 32310364-32310364 on Build GRCh38
rs750891293 Chr.20: 32310231-32310231 on Build GRCh38
rs746925417 Chr.20: 32310284-32310284 on Build GRCh38
rs760697145 Chr.20: 32310253-32310253 on Build GRCh38
rs373048849 Chr.20: 32310283-32310283 on Build GRCh38
rs202014684 Chr.20: 32310207-32310207 on Build GRCh38
rs983286168 Chr.20: 32310334-32310334 on Build GRCh38
rs952000297 Chr.20: 32310297-32310297 on Build GRCh38
rs761218320 Chr.20: 32310386-32310386 on Build GRCh38
rs758156143 Chr.20: 32310282-32310282 on Build GRCh38
rs1009081559 Chr.20: 32310205-32310208 on Build GRCh38
rs766448828 Chr.20: 32310259-32310259 on Build GRCh38
rs926946845 Chr.20: 32310416-32310416 on Build GRCh38
rs767921491 Chr.20: 32310219-32310219 on Build GRCh38
rs907685081 Chr.20: 32310365-32310365 on Build GRCh38
rs377401733 Chr.20: 32310387-32310387 on Build GRCh38
rs1027730832 Chr.20: 32310272-32310272 on Build GRCh38
rs964649308 Chr.20: 32310243-32310243 on Build GRCh38
rs752548410 Chr.20: 32310279-32310279 on Build GRCh38
rs778921981 Chr.20: 32310275-32310275 on Build GRCh38
rs773629692 Chr.20: 32310379-32310379 on Build GRCh38
rs759626291 Chr.20: 32310399-32310399 on Build GRCh38
rs1013907685 Chr.20: 32310336-32310336 on Build GRCh38
rs540409816 Chr.20: 32310317-32310317 on Build GRCh38
rs770388657 Chr.20: 32310299-32310299 on Build GRCh38
rs755234803 Chr.20: 32310270-32310270 on Build GRCh38
rs769441232 Chr.20: 32310328-32310328 on Build GRCh38
rs765572997 Chr.20: 32310404-32310404 on Build GRCh38

View Full Product Details