Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54569111-54569368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDE1B
Entrez Gene ID
5153
Gene Name
phosphodiesterase 1B
Gene Aliases
HEL-S-79p, PDE1B1, PDES1B
UniGene
Hs.530871
Gene Chromosome Location
Chr.12: 54549393-54579239 on Build GRCh38
Transcripts
NM_000924.3 NM_001165975.2 NM_001288768.1 NM_001288769.1 NM_001315534.1 NM_001315535.1 XM_011538456.1 XM_017019432.1 XM_017019433.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6049267 c.234G>A p.R78R Chr.12: 54569250-54569250 on Build GRCh38
1362821 c.277G>A p.V93M Chr.12: 54569233-54569233 on Build GRCh38
6859086 c.233G>A p.R78Q Chr.12: 54569249-54569249 on Build GRCh38
276689 c.228G>A p.R76R Chr.12: 54569184-54569184 on Build GRCh38
5434643 c.283C>T p.R95* Chr.12: 54569299-54569299 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs959313183 Chr.12: 54569323-54569323 on Build GRCh38
rs377170220 Chr.12: 54569309-54569309 on Build GRCh38
rs752442937 Chr.12: 54569261-54569261 on Build GRCh38
rs773362770 Chr.12: 54569322-54569322 on Build GRCh38
rs372746803 Chr.12: 54569250-54569250 on Build GRCh38
rs772266461 Chr.12: 54569314-54569314 on Build GRCh38
rs753650018 Chr.12: 54569247-54569247 on Build GRCh38
rs746223610 Chr.12: 54569271-54569271 on Build GRCh38
rs748432256 Chr.12: 54569302-54569302 on Build GRCh38
rs775771749 Chr.12: 54569140-54569140 on Build GRCh38
rs575814943 Chr.12: 54569337-54569337 on Build GRCh38
rs772982320 Chr.12: 54569212-54569212 on Build GRCh38
rs768806342 Chr.12: 54569295-54569295 on Build GRCh38
rs558783630 Chr.12: 54569340-54569340 on Build GRCh38
rs758093693 Chr.12: 54569265-54569265 on Build GRCh38
rs780617479 Chr.12: 54569275-54569275 on Build GRCh38
rs761895551 Chr.12: 54569205-54569205 on Build GRCh38
rs371717266 Chr.12: 54569198-54569198 on Build GRCh38
rs774670191 Chr.12: 54569203-54569203 on Build GRCh38
rs142799171 Chr.12: 54569249-54569249 on Build GRCh38
rs756491757 Chr.12: 54569274-54569274 on Build GRCh38
rs749723201 Chr.12: 54569284-54569284 on Build GRCh38
rs781732608 Chr.12: 54569266-54569266 on Build GRCh38
rs761908089 Chr.12: 54569287-54569287 on Build GRCh38
rs541950847 Chr.12: 54569218-54569218 on Build GRCh38
rs1015977531 Chr.12: 54569229-54569229 on Build GRCh38
rs772488653 Chr.12: 54569281-54569282 on Build GRCh38
rs574838194 Chr.12: 54569199-54569199 on Build GRCh38
rs535185955 Chr.12: 54569232-54569232 on Build GRCh38
rs1048627602 Chr.12: 54569164-54569164 on Build GRCh38
rs989268188 Chr.12: 54569338-54569338 on Build GRCh38
rs767337032 Chr.12: 54569208-54569208 on Build GRCh38
rs374242340 Chr.12: 54569296-54569296 on Build GRCh38
rs948593392 Chr.12: 54569282-54569282 on Build GRCh38

View Full Product Details