Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 100307-100578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPRL3
Entrez Gene ID
8131
Gene Name
NPR3 like, GATOR1 complex subunit
Gene Aliases
C16orf35, CGTHBA, HS-40, MARE, NPR3, RMD11
UniGene
Hs.19699
Gene Chromosome Location
Chr.16: 85801-138698 on Build GRCh38
Transcripts
NM_001039476.2 NM_001077350.2 NM_001243247.1 NM_001243248.1 NM_001243249.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2126436 c.668G>A p.S223N Chr.16: 100471-100471 on Build GRCh38
5363210 c.630-6_630-5delCT p.? Chr.16: 100514-100515 on Build GRCh38
5044403 c.700A>G p.K234E Chr.16: 100439-100439 on Build GRCh38
86086 c.726G>A p.L242L Chr.16: 100413-100413 on Build GRCh38
1217743 c.746A>C p.E249A Chr.16: 100393-100393 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759881682 Chr.16: 100343-100343 on Build GRCh38
rs749118108 Chr.16: 100374-100374 on Build GRCh38
rs761366554 Chr.16: 100426-100426 on Build GRCh38
rs201957188 Chr.16: 100487-100487 on Build GRCh38
rs374656133 Chr.16: 100518-100518 on Build GRCh38
rs112754051 Chr.16: 100353-100353 on Build GRCh38
rs763479734 Chr.16: 100405-100405 on Build GRCh38
rs772274896 Chr.16: 100379-100379 on Build GRCh38
rs760288230 Chr.16: 100524-100524 on Build GRCh38
rs747035776 Chr.16: 100384-100384 on Build GRCh38
rs774096708 Chr.16: 100506-100506 on Build GRCh38
rs916257624 Chr.16: 100387-100387 on Build GRCh38
rs377414615 Chr.16: 100354-100354 on Build GRCh38
rs766171956 Chr.16: 100525-100525 on Build GRCh38
rs766860772 Chr.16: 100328-100328 on Build GRCh38
rs556232826 Chr.16: 100498-100498 on Build GRCh38
rs755889116 Chr.16: 100372-100372 on Build GRCh38
rs201341279 Chr.16: 100486-100486 on Build GRCh38
rs200041907 Chr.16: 100394-100394 on Build GRCh38
rs781116168 Chr.16: 100364-100364 on Build GRCh38
rs763341816 Chr.16: 100502-100502 on Build GRCh38
rs749893964 Chr.16: 100523-100523 on Build GRCh38
rs769173039 Chr.16: 100503-100503 on Build GRCh38
rs940715864 Chr.16: 100358-100358 on Build GRCh38
rs554325689 Chr.16: 100383-100383 on Build GRCh38
rs753781901 Chr.16: 100528-100528 on Build GRCh38
rs770198792 Chr.16: 100500-100500 on Build GRCh38
rs756839067 Chr.16: 100360-100360 on Build GRCh38
rs766958453 Chr.16: 100434-100434 on Build GRCh38
rs776199294 Chr.16: 100501-100501 on Build GRCh38
rs745822946 Chr.16: 100366-100366 on Build GRCh38
rs368182847 Chr.16: 100497-100497 on Build GRCh38
rs752537537 Chr.16: 100543-100543 on Build GRCh38
rs765556171 Chr.16: 100348-100348 on Build GRCh38
rs763779646 Chr.16: 100406-100406 on Build GRCh38
rs747266609 Chr.16: 100493-100493 on Build GRCh38
rs754738681 Chr.16: 100331-100331 on Build GRCh38
rs367704264 Chr.16: 100534-100534 on Build GRCh38
rs532618698 Chr.16: 100521-100521 on Build GRCh38
rs368272925 Chr.16: 100443-100443 on Build GRCh38
rs577935445 Chr.16: 100391-100391 on Build GRCh38
rs947759739 Chr.16: 100390-100390 on Build GRCh38
rs754732050 Chr.16: 100532-100532 on Build GRCh38
rs779688768 Chr.16: 100373-100373 on Build GRCh38
rs770998374 Chr.16: 100494-100494 on Build GRCh38
rs770924192 Chr.16: 100386-100386 on Build GRCh38
rs751260495 Chr.16: 100425-100425 on Build GRCh38
rs777021343 Chr.16: 100340-100340 on Build GRCh38
rs375592476 Chr.16: 100395-100395 on Build GRCh38
rs780102496 Chr.16: 100477-100477 on Build GRCh38
rs1040446703 Chr.16: 100428-100428 on Build GRCh38
rs766032794 Chr.16: 100337-100337 on Build GRCh38
rs561515962 Chr.16: 100479-100479 on Build GRCh38
rs775687534 Chr.16: 100404-100404 on Build GRCh38

View Full Product Details