Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 180949-181210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HBQ1
Entrez Gene ID
3049
Gene Name
hemoglobin subunit theta 1
Gene Aliases
HBQ
UniGene
Hs.247921
Gene Chromosome Location
Chr.16: 180334-181179 on Build GRCh38
Transcripts
NM_005331.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1323701 c.392G>A p.S131N Chr.16: 181071-181071 on Build GRCh38
5475848 c.301-7G>A p.? Chr.16: 180973-180973 on Build GRCh38
7161150 c.374C>T p.S125L Chr.16: 181053-181053 on Build GRCh38
5548416 c.331G>A p.A111T Chr.16: 181010-181010 on Build GRCh38
7280564 c.312C>T p.H104H Chr.16: 180991-180991 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201357046 Chr.16: 180982-180982 on Build GRCh38
rs370841287 Chr.16: 180999-180999 on Build GRCh38
rs750864268 Chr.16: 181039-181039 on Build GRCh38
rs781650511 Chr.16: 180969-180969 on Build GRCh38
rs746138536 Chr.16: 180990-180991 on Build GRCh38
rs778448166 Chr.16: 181073-181073 on Build GRCh38
rs772116217 Chr.16: 181086-181086 on Build GRCh38
rs3929997 Chr.16: 181168-181168 on Build GRCh38
rs773118421 Chr.16: 181018-181018 on Build GRCh38
rs200446314 Chr.16: 181024-181024 on Build GRCh38
rs150819141 Chr.16: 181110-181110 on Build GRCh38
rs202203183 Chr.16: 181093-181093 on Build GRCh38
rs761040235 Chr.16: 181119-181120 on Build GRCh38
rs201426876 Chr.16: 181120-181120 on Build GRCh38
rs750707460 Chr.16: 181159-181159 on Build GRCh38
rs144961211 Chr.16: 181022-181022 on Build GRCh38
rs140989349 Chr.16: 180993-180993 on Build GRCh38
rs767757363 Chr.16: 181036-181036 on Build GRCh38
rs968153368 Chr.16: 181141-181141 on Build GRCh38
rs772305269 Chr.16: 180991-180997 on Build GRCh38
rs771741906 Chr.16: 181010-181010 on Build GRCh38
rs954347219 Chr.16: 181189-181189 on Build GRCh38
rs748812361 Chr.16: 180985-180985 on Build GRCh38
rs887321542 Chr.16: 181164-181164 on Build GRCh38
rs743861 Chr.16: 180975-180975 on Build GRCh38
rs775622709 Chr.16: 181113-181114 on Build GRCh38
rs775885863 Chr.16: 181122-181122 on Build GRCh38
rs912263955 Chr.16: 180974-180974 on Build GRCh38
rs559899613 Chr.16: 181013-181013 on Build GRCh38
rs748161655 Chr.16: 180998-180998 on Build GRCh38
rs374771812 Chr.16: 181126-181126 on Build GRCh38
rs567587801 Chr.16: 181178-181178 on Build GRCh38
rs200215753 Chr.16: 181133-181133 on Build GRCh38
rs779705914 Chr.16: 181055-181055 on Build GRCh38
rs931162647 Chr.16: 180997-180997 on Build GRCh38
rs745483593 Chr.16: 180970-180970 on Build GRCh38
rs768072211 Chr.16: 181157-181157 on Build GRCh38
rs1002823969 Chr.16: 181088-181088 on Build GRCh38
rs755653460 Chr.16: 181041-181041 on Build GRCh38
rs111543239 Chr.16: 181150-181150 on Build GRCh38
rs754457700 Chr.16: 181062-181062 on Build GRCh38
rs763303218 Chr.16: 181124-181124 on Build GRCh38
rs905676214 Chr.16: 181067-181067 on Build GRCh38
rs775651161 Chr.16: 181023-181023 on Build GRCh38
rs778856991 Chr.16: 180994-180994 on Build GRCh38
rs1021062352 Chr.16: 181140-181140 on Build GRCh38
rs748023581 Chr.16: 181076-181076 on Build GRCh38
rs752081969 Chr.16: 181027-181027 on Build GRCh38
rs1057482464 Chr.16: 181191-181191 on Build GRCh38

View Full Product Details