Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 145912389-145912134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEX11B
Entrez Gene ID
8799
Gene Name
peroxisomal biogenesis factor 11 beta
Gene Aliases
PEX11-BETA, PEX14B
UniGene
Hs.714608
Gene Chromosome Location
Chr.1: 145911348-145918924 on Build GRCh38
Transcripts
NM_001184795.1 NM_003846.2 NR_073491.1 NR_073492.1 NR_073493.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6744090 c.669C>A p.D223E Chr.1: 145912272-145912272 on Build GRCh38
3862586 c.725C>T p.S242F Chr.1: 145912216-145912216 on Build GRCh38
895186 c.649G>A p.D217N Chr.1: 145912292-145912292 on Build GRCh38
5377496 c.657C>T p.F219F Chr.1: 145912284-145912284 on Build GRCh38
895185 c.630C>T p.D210D Chr.1: 145912311-145912311 on Build GRCh38
View More

View Full Product Details