Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 85318-85577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPG
Entrez Gene ID
4350
Gene Name
N-methylpurine DNA glycosylase
Gene Aliases
AAG, ADPG, APNG, CRA36.1, MDG, Mid1, PIG11, PIG16, anpg
UniGene
Hs.459596
Gene Chromosome Location
Chr.16: 77019-85851 on Build GRCh38
Transcripts
NM_001015052.2 NM_001015054.2 NM_002434.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2143478 c.544C>T p.R182* Chr.16: 85424-85424 on Build GRCh38
6714653 c.545G>A p.R182Q Chr.16: 85425-85425 on Build GRCh38
2143480 c.635G>A p.R212H Chr.16: 85515-85515 on Build GRCh38
5850182 c.637G>A p.E213K Chr.16: 85517-85517 on Build GRCh38
1257900 c.589C>T p.R197C Chr.16: 85469-85469 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762883064 Chr.16: 85508-85508 on Build GRCh38
rs143541045 Chr.16: 85488-85489 on Build GRCh38
rs769191 Chr.16: 85477-85477 on Build GRCh38
rs200083505 Chr.16: 85368-85368 on Build GRCh38
rs575216465 Chr.16: 85492-85492 on Build GRCh38
rs893064586 Chr.16: 85457-85457 on Build GRCh38
rs750303980 Chr.16: 85382-85382 on Build GRCh38
rs940988565 Chr.16: 85376-85376 on Build GRCh38
rs746338668 Chr.16: 85408-85408 on Build GRCh38
rs907071937 Chr.16: 85424-85424 on Build GRCh38
rs771526808 Chr.16: 85442-85442 on Build GRCh38
rs755493706 Chr.16: 85519-85519 on Build GRCh38
rs761446491 Chr.16: 85424-85424 on Build GRCh38
rs781452593 Chr.16: 85407-85407 on Build GRCh38
rs142373562 Chr.16: 85515-85515 on Build GRCh38
rs749613799 Chr.16: 85414-85414 on Build GRCh38
rs558090660 Chr.16: 85543-85543 on Build GRCh38
rs776373615 Chr.16: 85499-85499 on Build GRCh38
rs754858141 Chr.16: 85399-85399 on Build GRCh38
rs757340771 Chr.16: 85406-85406 on Build GRCh38
rs974111690 Chr.16: 85354-85354 on Build GRCh38
rs776228234 Chr.16: 85551-85551 on Build GRCh38
rs771688051 Chr.16: 85487-85487 on Build GRCh38
rs200919197 Chr.16: 85481-85481 on Build GRCh38
rs145824088 Chr.16: 85415-85415 on Build GRCh38
rs774177528 Chr.16: 85396-85396 on Build GRCh38
rs779276542 Chr.16: 85482-85482 on Build GRCh38
rs375910503 Chr.16: 85391-85391 on Build GRCh38
rs992700321 Chr.16: 85542-85542 on Build GRCh38
rs748478874 Chr.16: 85533-85533 on Build GRCh38
rs765075901 Chr.16: 85501-85501 on Build GRCh38
rs903855275 Chr.16: 85468-85468 on Build GRCh38
rs767831098 Chr.16: 85471-85471 on Build GRCh38
rs747746976 Chr.16: 85483-85483 on Build GRCh38
rs534859796 Chr.16: 85465-85465 on Build GRCh38
rs772617796 Chr.16: 85445-85445 on Build GRCh38
rs570538336 Chr.16: 85460-85460 on Build GRCh38
rs765052101 Chr.16: 85477-85479 on Build GRCh38
rs754300245 Chr.16: 85518-85518 on Build GRCh38
rs541690579 Chr.16: 85546-85546 on Build GRCh38
rs766487967 Chr.16: 85517-85517 on Build GRCh38
rs775405600 Chr.16: 85505-85505 on Build GRCh38
rs201541383 Chr.16: 85365-85365 on Build GRCh38
rs577355454 Chr.16: 85536-85536 on Build GRCh38
rs369722201 Chr.16: 85392-85392 on Build GRCh38
rs750441127 Chr.16: 85472-85472 on Build GRCh38
rs1050764 Chr.16: 85522-85522 on Build GRCh38
rs773939189 Chr.16: 85417-85417 on Build GRCh38
rs767632759 Chr.16: 85514-85514 on Build GRCh38
rs759578895 Chr.16: 85500-85500 on Build GRCh38
rs552846697 Chr.16: 85469-85469 on Build GRCh38
rs934212442 Chr.16: 85339-85339 on Build GRCh38
rs770180276 Chr.16: 85411-85411 on Build GRCh38
rs547608974 Chr.16: 85375-85375 on Build GRCh38
rs983070483 Chr.16: 85347-85347 on Build GRCh38
rs867870089 Chr.16: 85498-85498 on Build GRCh38
rs766194520 Chr.16: 85458-85458 on Build GRCh38
rs200202323 Chr.16: 85516-85516 on Build GRCh38
rs761456411 Chr.16: 85425-85425 on Build GRCh38
rs535370395 Chr.16: 85479-85479 on Build GRCh38
rs746686232 Chr.16: 85541-85541 on Build GRCh38
rs776400933 Chr.16: 85399-85400 on Build GRCh38
rs780654126 Chr.16: 85412-85412 on Build GRCh38
rs777276807 Chr.16: 85538-85538 on Build GRCh38
rs200772943 Chr.16: 85524-85524 on Build GRCh38
rs774059355 Chr.16: 85363-85363 on Build GRCh38
rs751887856 Chr.16: 85405-85405 on Build GRCh38
rs147661014 Chr.16: 85493-85493 on Build GRCh38
rs760278233 Chr.16: 85456-85456 on Build GRCh38
rs757576353 Chr.16: 85470-85470 on Build GRCh38
rs998728803 Chr.16: 85444-85444 on Build GRCh38
rs148576448 Chr.16: 85461-85461 on Build GRCh38

View Full Product Details