Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104801336-104801573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB42
Entrez Gene ID
100128927
Gene Name
zinc finger and BTB domain containing 42
Gene Aliases
LCCS6, ZNF925
UniGene
Hs.170853
Gene Chromosome Location
Chr.14: 104800597-104804712 on Build GRCh38
Transcripts
NM_001137601.2 XM_017020911.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6346424 c.265G>T p.D89Y Chr.14: 104801462-104801462 on Build GRCh38
4776746 c.286G>A p.E96K Chr.14: 104801483-104801483 on Build GRCh38
5548317 c.298G>C p.A100P Chr.14: 104801495-104801495 on Build GRCh38
4620677 c.271C>T p.R91C Chr.14: 104801468-104801468 on Build GRCh38
3689975 c.176G>A p.G59D Chr.14: 104801373-104801373 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919005161 Chr.14: 104801420-104801420 on Build GRCh38
rs778382891 Chr.14: 104801499-104801499 on Build GRCh38
rs955933007 Chr.14: 104801402-104801402 on Build GRCh38
rs985139263 Chr.14: 104801517-104801517 on Build GRCh38
rs555761454 Chr.14: 104801464-104801464 on Build GRCh38
rs991583320 Chr.14: 104801373-104801373 on Build GRCh38
rs775815570 Chr.14: 104801441-104801441 on Build GRCh38
rs556354341 Chr.14: 104801365-104801365 on Build GRCh38
rs762464135 Chr.14: 104801520-104801520 on Build GRCh38
rs191737902 Chr.14: 104801383-104801383 on Build GRCh38
rs1022892503 Chr.14: 104801368-104801368 on Build GRCh38
rs570148369 Chr.14: 104801378-104801378 on Build GRCh38
rs981286554 Chr.14: 104801390-104801390 on Build GRCh38
rs750004064 Chr.14: 104801536-104801536 on Build GRCh38
rs1031474075 Chr.14: 104801393-104801393 on Build GRCh38
rs756663629 Chr.14: 104801434-104801434 on Build GRCh38
rs1033343666 Chr.14: 104801363-104801363 on Build GRCh38
rs932382238 Chr.14: 104801514-104801514 on Build GRCh38
rs572006913 Chr.14: 104801494-104801494 on Build GRCh38
rs887840252 Chr.14: 104801364-104801364 on Build GRCh38
rs943609957 Chr.14: 104801506-104801506 on Build GRCh38
rs1049196131 Chr.14: 104801450-104801450 on Build GRCh38
rs763081051 Chr.14: 104801457-104801457 on Build GRCh38

View Full Product Details