Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54593878-54594119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT11
Entrez Gene ID
26027
Gene Name
acyl-CoA thioesterase 11
Gene Aliases
BFIT, STARD14, THEA, THEM1
UniGene
Hs.745173
Gene Chromosome Location
Chr.1: 54548134-54634744 on Build GRCh38
Transcripts
NM_015547.3 NM_147161.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
426476 c.457C>G p.R153G Chr.1: 54594025-54594025 on Build GRCh38
426474 c.430A>G p.K144E Chr.1: 54593998-54593998 on Build GRCh38
5780668 c.454C>G p.R152G Chr.1: 54594022-54594022 on Build GRCh38
3944177 c.442A>C p.T148P Chr.1: 54594010-54594010 on Build GRCh38
4546961 c.411G>A p.E137E Chr.1: 54593979-54593979 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775100884 Chr.1: 54594024-54594024 on Build GRCh38
rs758770244 Chr.1: 54594079-54594079 on Build GRCh38
rs1018602412 Chr.1: 54594063-54594063 on Build GRCh38
rs963757473 Chr.1: 54594082-54594083 on Build GRCh38
rs1057095419 Chr.1: 54593900-54593900 on Build GRCh38
rs780870855 Chr.1: 54594016-54594016 on Build GRCh38
rs150766970 Chr.1: 54593916-54593916 on Build GRCh38
rs772387844 Chr.1: 54594015-54594015 on Build GRCh38
rs761663984 Chr.1: 54594033-54594033 on Build GRCh38
rs2304307 Chr.1: 54593920-54593920 on Build GRCh38
rs762558370 Chr.1: 54594025-54594025 on Build GRCh38
rs769594892 Chr.1: 54594023-54594023 on Build GRCh38
rs184760562 Chr.1: 54594051-54594051 on Build GRCh38
rs755436167 Chr.1: 54594002-54594002 on Build GRCh38
rs564247882 Chr.1: 54594067-54594067 on Build GRCh38
rs559815271 Chr.1: 54593967-54593967 on Build GRCh38
rs112888615 Chr.1: 54593927-54593927 on Build GRCh38
rs755023769 Chr.1: 54594068-54594068 on Build GRCh38
rs767765385 Chr.1: 54593951-54593951 on Build GRCh38
rs769674051 Chr.1: 54593983-54593983 on Build GRCh38
rs757017367 Chr.1: 54593946-54593946 on Build GRCh38
rs370568235 Chr.1: 54594009-54594009 on Build GRCh38
rs79023440 Chr.1: 54594099-54594099 on Build GRCh38
rs35306628 Chr.1: 54594009-54594009 on Build GRCh38
rs377602880 Chr.1: 54593940-54593940 on Build GRCh38
rs986773000 Chr.1: 54594058-54594058 on Build GRCh38
rs549768266 Chr.1: 54594087-54594087 on Build GRCh38
rs980687516 Chr.1: 54594085-54594085 on Build GRCh38
rs953963433 Chr.1: 54594017-54594017 on Build GRCh38
rs140453182 Chr.1: 54593960-54593960 on Build GRCh38
rs144741986 Chr.1: 54594052-54594052 on Build GRCh38
rs748484488 Chr.1: 54594010-54594010 on Build GRCh38
rs533155718 Chr.1: 54594081-54594081 on Build GRCh38
rs780155096 Chr.1: 54593961-54593961 on Build GRCh38
rs745566637 Chr.1: 54594022-54594022 on Build GRCh38
rs750710661 Chr.1: 54593958-54593958 on Build GRCh38
rs762936488 Chr.1: 54593919-54593919 on Build GRCh38
rs569924055 Chr.1: 54594094-54594094 on Build GRCh38
rs768581245 Chr.1: 54594026-54594026 on Build GRCh38
rs759535791 Chr.1: 54593967-54593967 on Build GRCh38
rs775548801 Chr.1: 54593901-54593901 on Build GRCh38
rs767254660 Chr.1: 54594044-54594044 on Build GRCh38
rs368400152 Chr.1: 54594056-54594056 on Build GRCh38
rs774212254 Chr.1: 54594031-54594031 on Build GRCh38
rs767326119 Chr.1: 54594059-54594059 on Build GRCh38
rs927478744 Chr.1: 54594098-54594098 on Build GRCh38
rs147894114 Chr.1: 54594083-54594083 on Build GRCh38
rs751512271 Chr.1: 54593912-54593913 on Build GRCh38
rs753188654 Chr.1: 54594074-54594074 on Build GRCh38

View Full Product Details