Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48826142-48826415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNB3
Entrez Gene ID
784
Gene Name
calcium voltage-gated channel auxiliary subunit beta 3
Gene Aliases
CAB3, CACNLB3
UniGene
Hs.250712
Gene Chromosome Location
Chr.12: 48814432-48828943 on Build GRCh38
Transcripts
NM_000725.3 NM_001206915.1 NM_001206916.1 NM_001206917.1 XM_011538727.2 XM_011538728.2 XM_017019957.1 XM_017019958.1 XM_017019959.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
939903 c.767G>A p.R256H Chr.12: 48826391-48826391 on Build GRCh38
4430640 c.751C>T p.Q251* Chr.12: 48826375-48826375 on Build GRCh38
939904 c.764G>A p.R255H Chr.12: 48826391-48826391 on Build GRCh38
279528 c.743C>T p.A248V Chr.12: 48826367-48826367 on Build GRCh38
4430641 c.748C>T p.Q250* Chr.12: 48826375-48826375 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751873915 Chr.12: 48826385-48826385 on Build GRCh38
rs975190383 Chr.12: 48826284-48826284 on Build GRCh38
rs1011558769 Chr.12: 48826260-48826260 on Build GRCh38
rs756553313 Chr.12: 48826323-48826323 on Build GRCh38
rs1030089272 Chr.12: 48826192-48826192 on Build GRCh38
rs758647170 Chr.12: 48826367-48826367 on Build GRCh38
rs773148613 Chr.12: 48826376-48826387 on Build GRCh38
rs547372189 Chr.12: 48826221-48826221 on Build GRCh38
rs962921569 Chr.12: 48826292-48826292 on Build GRCh38
rs10875878 Chr.12: 48826249-48826249 on Build GRCh38
rs76025846 Chr.12: 48826191-48826191 on Build GRCh38
rs767395204 Chr.12: 48826331-48826331 on Build GRCh38
rs765165048 Chr.12: 48826348-48826348 on Build GRCh38
rs750314856 Chr.12: 48826332-48826332 on Build GRCh38
rs377762667 Chr.12: 48826355-48826355 on Build GRCh38
rs1021551005 Chr.12: 48826274-48826274 on Build GRCh38
rs964782363 Chr.12: 48826282-48826282 on Build GRCh38
rs767270545 Chr.12: 48826184-48826184 on Build GRCh38
rs906990831 Chr.12: 48826189-48826189 on Build GRCh38
rs778175847 Chr.12: 48826368-48826368 on Build GRCh38
rs781661766 Chr.12: 48826391-48826391 on Build GRCh38
rs985846358 Chr.12: 48826248-48826248 on Build GRCh38
rs972900012 Chr.12: 48826386-48826386 on Build GRCh38
rs757670331 Chr.12: 48826388-48826388 on Build GRCh38
rs948423178 Chr.12: 48826169-48826169 on Build GRCh38
rs183144910 Chr.12: 48826181-48826181 on Build GRCh38
rs760734741 Chr.12: 48826345-48826345 on Build GRCh38
rs747727347 Chr.12: 48826200-48826200 on Build GRCh38
rs752840438 Chr.12: 48826359-48826359 on Build GRCh38

View Full Product Details