Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 130679306-130679531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIK3R4
Entrez Gene ID
30849
Gene Name
phosphoinositide-3-kinase regulatory subunit 4
Gene Aliases
VPS15, p150
UniGene
Hs.149032
Gene Chromosome Location
Chr.3: 130678934-130746852 on Build GRCh38
Transcripts
NM_014602.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6163188 c.4026G>T p.Q1342H Chr.3: 130679366-130679366 on Build GRCh38
1419014 c.4041T>C p.T1347T Chr.3: 130679351-130679351 on Build GRCh38
6511539 c.4027G>T p.G1343C Chr.3: 130679365-130679365 on Build GRCh38
1693456 c.3968C>T p.S1323F Chr.3: 130679424-130679424 on Build GRCh38
1327878 c.3927A>C p.K1309N Chr.3: 130679465-130679465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765217350 Chr.3: 130679440-130679440 on Build GRCh38
rs537201013 Chr.3: 130679444-130679444 on Build GRCh38
rs767497266 Chr.3: 130679493-130679494 on Build GRCh38
rs199874599 Chr.3: 130679423-130679423 on Build GRCh38
rs111650637 Chr.3: 130679454-130679454 on Build GRCh38
rs531376166 Chr.3: 130679385-130679385 on Build GRCh38
rs758176708 Chr.3: 130679377-130679377 on Build GRCh38
rs1041461896 Chr.3: 130679507-130679507 on Build GRCh38
rs869222604 Chr.3: 130679433-130679433 on Build GRCh38
rs958315137 Chr.3: 130679339-130679339 on Build GRCh38
rs766122644 Chr.3: 130679344-130679345 on Build GRCh38
rs753838129 Chr.3: 130679349-130679349 on Build GRCh38
rs892897152 Chr.3: 130679460-130679460 on Build GRCh38
rs774953971 Chr.3: 130679378-130679378 on Build GRCh38
rs200858388 Chr.3: 130679369-130679369 on Build GRCh38
rs766514140 Chr.3: 130679426-130679426 on Build GRCh38
rs140737482 Chr.3: 130679468-130679468 on Build GRCh38
rs370833096 Chr.3: 130679397-130679397 on Build GRCh38
rs752537591 Chr.3: 130679375-130679375 on Build GRCh38
rs758193124 Chr.3: 130679445-130679445 on Build GRCh38
rs766315043 Chr.3: 130679340-130679340 on Build GRCh38
rs145121897 Chr.3: 130679402-130679402 on Build GRCh38
rs759060722 Chr.3: 130679486-130679487 on Build GRCh38
rs777548732 Chr.3: 130679383-130679383 on Build GRCh38
rs201481225 Chr.3: 130679336-130679336 on Build GRCh38
rs751245059 Chr.3: 130679455-130679455 on Build GRCh38
rs769891029 Chr.3: 130679394-130679395 on Build GRCh38
rs773501007 Chr.3: 130679418-130679418 on Build GRCh38
rs769324167 Chr.3: 130679399-130679399 on Build GRCh38
rs748594594 Chr.3: 130679406-130679406 on Build GRCh38
rs550061438 Chr.3: 130679417-130679417 on Build GRCh38
rs538802327 Chr.3: 130679433-130679433 on Build GRCh38
rs756817117 Chr.3: 130679458-130679458 on Build GRCh38
rs945749321 Chr.3: 130679491-130679491 on Build GRCh38
rs758913806 Chr.3: 130679386-130679386 on Build GRCh38
rs1036124027 Chr.3: 130679456-130679456 on Build GRCh38
rs914083942 Chr.3: 130679368-130679368 on Build GRCh38
rs780516891 Chr.3: 130679387-130679387 on Build GRCh38
rs759590880 Chr.3: 130679439-130679439 on Build GRCh38
rs140271304 Chr.3: 130679357-130679357 on Build GRCh38

View Full Product Details