Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102718652-102718877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD9
Entrez Gene ID
84804
Gene Name
major facilitator superfamily domain containing 9
Gene Aliases
-
UniGene
Hs.590898
Gene Chromosome Location
Chr.2: 102713867-102736888 on Build GRCh38
Transcripts
NM_001322080.1 NM_001322081.1 NM_032718.4 NR_136173.1 NR_136174.1 XM_011512005.2 XM_011512006.2 XM_011512007.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5881196 c.998C>T p.A333V Chr.2: 102718847-102718847 on Build GRCh38
328822 c.982A>T p.S328C Chr.2: 102718863-102718863 on Build GRCh38
1481804 c.1188G>T p.T396T Chr.2: 102718657-102718657 on Build GRCh38
5859799 c.1018G>A p.G340R Chr.2: 102718827-102718827 on Build GRCh38
6848992 c.986G>A p.S329N Chr.2: 102718859-102718859 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776881213 Chr.2: 102718785-102718785 on Build GRCh38
rs760735823 Chr.2: 102718707-102718707 on Build GRCh38
rs765310955 Chr.2: 102718790-102718790 on Build GRCh38
rs140655272 Chr.2: 102718756-102718756 on Build GRCh38
rs578146378 Chr.2: 102718846-102718846 on Build GRCh38
rs754504524 Chr.2: 102718815-102718815 on Build GRCh38
rs748502328 Chr.2: 102718762-102718762 on Build GRCh38
rs770301367 Chr.2: 102718674-102718674 on Build GRCh38
rs370598161 Chr.2: 102718794-102718794 on Build GRCh38
rs754417483 Chr.2: 102718745-102718745 on Build GRCh38
rs758043502 Chr.2: 102718731-102718731 on Build GRCh38
rs748534014 Chr.2: 102718833-102718833 on Build GRCh38
rs142876545 Chr.2: 102718814-102718814 on Build GRCh38
rs377082803 Chr.2: 102718761-102718761 on Build GRCh38
rs768946691 Chr.2: 102718688-102718688 on Build GRCh38
rs17027809 Chr.2: 102718795-102718795 on Build GRCh38
rs753769125 Chr.2: 102718717-102718717 on Build GRCh38
rs148301225 Chr.2: 102718839-102718839 on Build GRCh38
rs538334164 Chr.2: 102718821-102718821 on Build GRCh38
rs776037723 Chr.2: 102718678-102718678 on Build GRCh38
rs752452824 Chr.2: 102718730-102718730 on Build GRCh38
rs566616874 Chr.2: 102718764-102718764 on Build GRCh38
rs971974327 Chr.2: 102718744-102718744 on Build GRCh38
rs749812297 Chr.2: 102718683-102718683 on Build GRCh38
rs79177698 Chr.2: 102718809-102718809 on Build GRCh38
rs1000198734 Chr.2: 102718725-102718725 on Build GRCh38
rs759729239 Chr.2: 102718851-102718851 on Build GRCh38
rs749985431 Chr.2: 102718818-102718818 on Build GRCh38
rs749420827 Chr.2: 102718686-102718688 on Build GRCh38
rs766403696 Chr.2: 102718708-102718708 on Build GRCh38
rs371519262 Chr.2: 102718698-102718698 on Build GRCh38
rs760963785 Chr.2: 102718771-102718771 on Build GRCh38
rs765084111 Chr.2: 102718724-102718724 on Build GRCh38
rs34096572 Chr.2: 102718719-102718719 on Build GRCh38
rs771189567 Chr.2: 102718708-102718710 on Build GRCh38
rs746546160 Chr.2: 102718670-102718670 on Build GRCh38
rs779356874 Chr.2: 102718831-102718831 on Build GRCh38
rs773361899 Chr.2: 102718765-102718765 on Build GRCh38
rs375902859 Chr.2: 102718848-102718848 on Build GRCh38
rs774733342 Chr.2: 102718691-102718691 on Build GRCh38
rs772265230 Chr.2: 102718838-102718838 on Build GRCh38
rs372604714 Chr.2: 102718752-102718752 on Build GRCh38
rs766498615 Chr.2: 102718777-102718777 on Build GRCh38
rs746522392 Chr.2: 102718742-102718742 on Build GRCh38
rs752502355 Chr.2: 102718793-102718793 on Build GRCh38
rs759493345 Chr.2: 102718788-102718788 on Build GRCh38
rs777328058 Chr.2: 102718741-102718741 on Build GRCh38
rs767682152 Chr.2: 102718701-102718701 on Build GRCh38
rs140585828 Chr.2: 102718845-102718845 on Build GRCh38
rs368670792 Chr.2: 102718813-102718813 on Build GRCh38
rs780687642 Chr.2: 102718755-102718755 on Build GRCh38
rs773313013 Chr.2: 102718704-102718704 on Build GRCh38
rs1016040243 Chr.2: 102718703-102718703 on Build GRCh38

View Full Product Details