Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 66442843-66443114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEIS1
Entrez Gene ID
4211
Gene Name
Meis homeobox 1
Gene Aliases
-
UniGene
Hs.526754
Gene Chromosome Location
Chr.2: 66435125-66572765 on Build GRCh38
Transcripts
NM_002398.2 XM_005264321.2 XM_005264322.2 XM_005264323.2 XM_005264324.4 XM_005264325.4 XM_017004141.1 XM_017004142.1 XM_017004143.1 XM_017004144.1 XM_017004145.1 XM_017004146.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6307517 c.512A>C p.H171P Chr.2: 66442930-66442930 on Build GRCh38
5354547 c.488A>C p.H163P Chr.2: 66442906-66442906 on Build GRCh38
5506685 c.524G>A p.S175N Chr.2: 66442942-66442942 on Build GRCh38
6256570 c.562G>T p.D188Y Chr.2: 66442980-66442980 on Build GRCh38
3055437 c.587_590delCAGA p.D197fs*4 Chr.2: 66443005-66443008 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762812768 Chr.2: 66442878-66442878 on Build GRCh38
rs750064744 Chr.2: 66442967-66442967 on Build GRCh38
rs372313599 Chr.2: 66443070-66443070 on Build GRCh38
rs777773287 Chr.2: 66443012-66443012 on Build GRCh38
rs552329583 Chr.2: 66443028-66443028 on Build GRCh38
rs368068335 Chr.2: 66443090-66443090 on Build GRCh38
rs560708954 Chr.2: 66442942-66442942 on Build GRCh38
rs763260544 Chr.2: 66442876-66442877 on Build GRCh38
rs554620476 Chr.2: 66443040-66443040 on Build GRCh38
rs369833361 Chr.2: 66443025-66443025 on Build GRCh38
rs199688276 Chr.2: 66442893-66442893 on Build GRCh38
rs781493613 Chr.2: 66443043-66443043 on Build GRCh38
rs757005566 Chr.2: 66443091-66443092 on Build GRCh38
rs764005971 Chr.2: 66443079-66443080 on Build GRCh38
rs1052923098 Chr.2: 66443009-66443009 on Build GRCh38
rs758451287 Chr.2: 66442987-66442987 on Build GRCh38
rs772211406 Chr.2: 66442978-66442978 on Build GRCh38
rs768606869 Chr.2: 66443052-66443052 on Build GRCh38
rs760717673 Chr.2: 66443047-66443047 on Build GRCh38
rs540467903 Chr.2: 66442907-66442907 on Build GRCh38
rs368074777 Chr.2: 66442880-66442880 on Build GRCh38
rs375137690 Chr.2: 66443071-66443071 on Build GRCh38
rs759602778 Chr.2: 66443080-66443080 on Build GRCh38
rs759852650 Chr.2: 66443092-66443092 on Build GRCh38
rs774494753 Chr.2: 66442988-66442990 on Build GRCh38
rs755348268 Chr.2: 66442983-66442983 on Build GRCh38
rs766315139 Chr.2: 66442883-66442884 on Build GRCh38
rs775405225 Chr.2: 66442982-66442982 on Build GRCh38
rs774454871 Chr.2: 66442883-66442883 on Build GRCh38
rs776367043 Chr.2: 66443090-66443090 on Build GRCh38
rs753823045 Chr.2: 66443088-66443089 on Build GRCh38
rs539953965 Chr.2: 66443067-66443067 on Build GRCh38
rs371255938 Chr.2: 66442895-66442895 on Build GRCh38
rs775218475 Chr.2: 66442877-66442877 on Build GRCh38
rs753232642 Chr.2: 66442986-66442986 on Build GRCh38
rs760299731 Chr.2: 66442968-66442968 on Build GRCh38
rs892646699 Chr.2: 66443022-66443022 on Build GRCh38
rs375874864 Chr.2: 66442985-66442985 on Build GRCh38
rs200317215 Chr.2: 66443054-66443054 on Build GRCh38
rs560234782 Chr.2: 66443080-66443080 on Build GRCh38

View Full Product Details