Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 103586604-103586837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCEAL4
Entrez Gene ID
79921
Gene Name
transcription elongation factor A like 4
Gene Aliases
NPD017, WEX7
UniGene
Hs.194329
Gene Chromosome Location
Chr.X: 103576231-103587736 on Build GRCh38
Transcripts
NM_001006935.2 NM_001006937.2 NM_001300901.1 NM_001305840.1 NM_001305841.1 NM_001305842.1 NM_024863.5 XM_017029856.1 XM_017029857.1

SNP Detail

SNP ID Chromosome Location
rs777326854 Chr.X: 103586710-103586710 on Build GRCh38
rs866893955 Chr.X: 103586740-103586741 on Build GRCh38
rs749300992 Chr.X: 103586764-103586764 on Build GRCh38
rs781042964 Chr.X: 103586736-103586738 on Build GRCh38
rs758735245 Chr.X: 103586736-103586736 on Build GRCh38
rs778839913 Chr.X: 103586766-103586766 on Build GRCh38
rs375212214 Chr.X: 103586660-103586660 on Build GRCh38
rs773508493 Chr.X: 103586673-103586673 on Build GRCh38
rs62621172 Chr.X: 103586652-103586652 on Build GRCh38
rs755664856 Chr.X: 103586746-103586746 on Build GRCh38
rs765701439 Chr.X: 103586730-103586730 on Build GRCh38
rs760859338 Chr.X: 103586691-103586691 on Build GRCh38
rs771942991 Chr.X: 103586789-103586789 on Build GRCh38
rs747212870 Chr.X: 103586807-103586807 on Build GRCh38
rs770727393 Chr.X: 103586724-103586730 on Build GRCh38
rs763014665 Chr.X: 103586735-103586735 on Build GRCh38
rs774001516 Chr.X: 103586659-103586659 on Build GRCh38
rs368081043 Chr.X: 103586644-103586644 on Build GRCh38
rs779513354 Chr.X: 103586753-103586753 on Build GRCh38
rs772187446 Chr.X: 103586665-103586665 on Build GRCh38
rs759947835 Chr.X: 103586711-103586711 on Build GRCh38
rs754865396 Chr.X: 103586765-103586765 on Build GRCh38
rs745397973 Chr.X: 103586636-103586636 on Build GRCh38
rs780759122 Chr.X: 103586632-103586632 on Build GRCh38
rs766525275 Chr.X: 103586709-103586709 on Build GRCh38
rs750042611 Chr.X: 103586741-103586741 on Build GRCh38
rs773598572 Chr.X: 103586791-103586791 on Build GRCh38
rs767181694 Chr.X: 103586739-103586739 on Build GRCh38
rs147553313 Chr.X: 103586781-103586781 on Build GRCh38
rs745873765 Chr.X: 103586783-103586783 on Build GRCh38
rs756857801 Chr.X: 103586630-103586630 on Build GRCh38

View Full Product Details