Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 19698535-19698808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TWISTNB
Entrez Gene ID
221830
Gene Name
TWIST neighbor
Gene Aliases
-
UniGene
Hs.353035
Gene Chromosome Location
Chr.7: 19695462-19709037 on Build GRCh38
Transcripts
NM_001002926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1187264 c.725G>T p.S242I Chr.7: 19698608-19698608 on Build GRCh38
248157 c.619C>T p.R207C Chr.7: 19698714-19698714 on Build GRCh38
3832459 c.615C>G p.F205L Chr.7: 19698718-19698718 on Build GRCh38
3307425 c.751G>A p.A251T Chr.7: 19698582-19698582 on Build GRCh38
3768310 c.696G>C p.K232N Chr.7: 19698637-19698637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140577942 Chr.7: 19698648-19698648 on Build GRCh38
rs1049437227 Chr.7: 19698691-19698691 on Build GRCh38
rs758624888 Chr.7: 19698631-19698631 on Build GRCh38
rs914329718 Chr.7: 19698608-19698608 on Build GRCh38
rs757393431 Chr.7: 19698773-19698773 on Build GRCh38
rs555342790 Chr.7: 19698581-19698581 on Build GRCh38
rs774898966 Chr.7: 19698713-19698713 on Build GRCh38
rs150404597 Chr.7: 19698700-19698700 on Build GRCh38
rs370526884 Chr.7: 19698615-19698615 on Build GRCh38
rs751641540 Chr.7: 19698755-19698755 on Build GRCh38
rs944981441 Chr.7: 19698717-19698717 on Build GRCh38
rs757772418 Chr.7: 19698639-19698639 on Build GRCh38
rs747041820 Chr.7: 19698637-19698637 on Build GRCh38
rs139060675 Chr.7: 19698666-19698666 on Build GRCh38
rs764243242 Chr.7: 19698753-19698753 on Build GRCh38
rs763558222 Chr.7: 19698561-19698561 on Build GRCh38
rs746367514 Chr.7: 19698680-19698680 on Build GRCh38
rs768317854 Chr.7: 19698695-19698695 on Build GRCh38
rs199908969 Chr.7: 19698737-19698737 on Build GRCh38
rs1007521771 Chr.7: 19698732-19698732 on Build GRCh38
rs763841038 Chr.7: 19698767-19698768 on Build GRCh38
rs549843354 Chr.7: 19698613-19698613 on Build GRCh38
rs761877350 Chr.7: 19698579-19698579 on Build GRCh38
rs999679899 Chr.7: 19698760-19698760 on Build GRCh38
rs763105859 Chr.7: 19698726-19698726 on Build GRCh38
rs146190540 Chr.7: 19698567-19698567 on Build GRCh38
rs777860794 Chr.7: 19698636-19698636 on Build GRCh38
rs752206207 Chr.7: 19698738-19698739 on Build GRCh38
rs898907857 Chr.7: 19698690-19698690 on Build GRCh38
rs990282026 Chr.7: 19698582-19698582 on Build GRCh38
rs774025015 Chr.7: 19698697-19698697 on Build GRCh38
rs748929480 Chr.7: 19698689-19698689 on Build GRCh38
rs1036095879 Chr.7: 19698688-19698688 on Build GRCh38
rs531318699 Chr.7: 19698587-19698587 on Build GRCh38
rs927773407 Chr.7: 19698677-19698677 on Build GRCh38
rs375352231 Chr.7: 19698714-19698714 on Build GRCh38
rs187171103 Chr.7: 19698619-19698619 on Build GRCh38
rs757999271 Chr.7: 19698744-19698744 on Build GRCh38
rs759030662 Chr.7: 19698647-19698655 on Build GRCh38
rs770112631 Chr.7: 19698686-19698686 on Build GRCh38
rs753639473 Chr.7: 19698716-19698716 on Build GRCh38
rs766193466 Chr.7: 19698616-19698616 on Build GRCh38

View Full Product Details