Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 74245090-74245363 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VSX2
Entrez Gene ID
338917
Gene Name
visual system homeobox 2
Gene Aliases
CHX10, HOX10, MCOP2, MCOPCB3, RET1
UniGene
Hs.449771
Gene Chromosome Location
Chr.14: 74239472-74262738 on Build GRCh38
Transcripts
NM_182894.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5805645 c.476A>T p.Q159L Chr.14: 74245185-74245185 on Build GRCh38
130182 c.493A>C p.K165Q Chr.14: 74245202-74245202 on Build GRCh38
5759229 c.578A>G p.Q193R Chr.14: 74245287-74245287 on Build GRCh38
698685 c.528T>C p.Y176Y Chr.14: 74245237-74245237 on Build GRCh38
1300821 c.481G>C p.E161Q Chr.14: 74245190-74245190 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147167593 Chr.14: 74245248-74245248 on Build GRCh38
rs551394732 Chr.14: 74245179-74245179 on Build GRCh38
rs201119252 Chr.14: 74245318-74245318 on Build GRCh38
rs78509400 Chr.14: 74245231-74245231 on Build GRCh38
rs113142527 Chr.14: 74245256-74245256 on Build GRCh38
rs767911905 Chr.14: 74245271-74245271 on Build GRCh38
rs765447479 Chr.14: 74245169-74245169 on Build GRCh38
rs1027968685 Chr.14: 74245311-74245311 on Build GRCh38
rs763144610 Chr.14: 74245177-74245177 on Build GRCh38
rs779358821 Chr.14: 74245309-74245310 on Build GRCh38
rs754236214 Chr.14: 74245277-74245277 on Build GRCh38
rs143366810 Chr.14: 74245192-74245192 on Build GRCh38
rs727504220 Chr.14: 74245285-74245285 on Build GRCh38
rs769645897 Chr.14: 74245313-74245313 on Build GRCh38
rs886050738 Chr.14: 74245242-74245242 on Build GRCh38
rs1022601308 Chr.14: 74245329-74245329 on Build GRCh38
rs1004966866 Chr.14: 74245247-74245247 on Build GRCh38
rs745654446 Chr.14: 74245312-74245312 on Build GRCh38
rs373577377 Chr.14: 74245125-74245125 on Build GRCh38
rs777105103 Chr.14: 74245294-74245294 on Build GRCh38
rs775779850 Chr.14: 74245253-74245253 on Build GRCh38
rs750768011 Chr.14: 74245272-74245272 on Build GRCh38
rs878900418 Chr.14: 74245202-74245202 on Build GRCh38
rs771439780 Chr.14: 74245300-74245302 on Build GRCh38
rs774267021 Chr.14: 74245332-74245332 on Build GRCh38
rs772944015 Chr.14: 74245148-74245148 on Build GRCh38
rs751830363 Chr.14: 74245263-74245263 on Build GRCh38
rs777493485 Chr.14: 74245232-74245232 on Build GRCh38
rs994603286 Chr.14: 74245205-74245205 on Build GRCh38
rs375663408 Chr.14: 74245226-74245226 on Build GRCh38
rs760519200 Chr.14: 74245154-74245154 on Build GRCh38
rs764023843 Chr.14: 74245261-74245261 on Build GRCh38
rs113575275 Chr.14: 74245331-74245331 on Build GRCh38
rs528252884 Chr.14: 74245147-74245147 on Build GRCh38
rs898972555 Chr.14: 74245199-74245199 on Build GRCh38
rs747842829 Chr.14: 74245133-74245133 on Build GRCh38
rs776731233 Chr.14: 74245159-74245159 on Build GRCh38
rs761903752 Chr.14: 74245264-74245264 on Build GRCh38
rs752801719 Chr.14: 74245172-74245172 on Build GRCh38
rs201354547 Chr.14: 74245273-74245273 on Build GRCh38
rs746590938 Chr.14: 74245240-74245240 on Build GRCh38
rs930892204 Chr.14: 74245201-74245201 on Build GRCh38
rs757655338 Chr.14: 74245184-74245184 on Build GRCh38
rs371028343 Chr.14: 74245325-74245325 on Build GRCh38
rs1051710907 Chr.14: 74245171-74245171 on Build GRCh38
rs573993188 Chr.14: 74245214-74245214 on Build GRCh38
rs550984628 Chr.14: 74245213-74245213 on Build GRCh38
rs759505373 Chr.14: 74245162-74245162 on Build GRCh38
rs746234051 Chr.14: 74245324-74245325 on Build GRCh38
rs770570869 Chr.14: 74245158-74245158 on Build GRCh38
rs182972044 Chr.14: 74245288-74245288 on Build GRCh38
rs539178171 Chr.14: 74245305-74245305 on Build GRCh38
rs750962175 Chr.14: 74245212-74245212 on Build GRCh38
rs35435463 Chr.14: 74245180-74245180 on Build GRCh38
rs947511976 Chr.14: 74245121-74245121 on Build GRCh38
rs559035140 Chr.14: 74245308-74245308 on Build GRCh38
rs143480909 Chr.14: 74245241-74245241 on Build GRCh38
rs748800493 Chr.14: 74245297-74245298 on Build GRCh38

View Full Product Details