Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 83917513-83917784 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTLL7
Entrez Gene ID
79739
Gene Name
tubulin tyrosine ligase like 7
Gene Aliases
-
UniGene
Hs.445826
Gene Chromosome Location
Chr.1: 83865021-83999340 on Build GRCh38
Transcripts
NM_024686.4 XM_005271208.2 XM_011542159.2 XM_017002347.1 XM_017002348.1 XM_017002349.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7177786 c.1576C>G p.R526G Chr.1: 83917615-83917615 on Build GRCh38
4990645 c.1501G>A p.E501K Chr.1: 83917690-83917690 on Build GRCh38
1230849 c.1507G>T p.D503Y Chr.1: 83917684-83917684 on Build GRCh38
3492913 c.1569C>T p.T523T Chr.1: 83917622-83917622 on Build GRCh38
248939 c.1576C>T p.R526* Chr.1: 83917615-83917615 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781080724 Chr.1: 83917615-83917615 on Build GRCh38
rs765638123 Chr.1: 83917593-83917593 on Build GRCh38
rs377052183 Chr.1: 83917601-83917601 on Build GRCh38
rs980214983 Chr.1: 83917756-83917756 on Build GRCh38
rs369067476 Chr.1: 83917559-83917559 on Build GRCh38
rs764527882 Chr.1: 83917739-83917739 on Build GRCh38
rs760100294 Chr.1: 83917703-83917703 on Build GRCh38
rs201237224 Chr.1: 83917631-83917631 on Build GRCh38
rs760972630 Chr.1: 83917738-83917739 on Build GRCh38
rs577416666 Chr.1: 83917623-83917623 on Build GRCh38
rs758820900 Chr.1: 83917599-83917599 on Build GRCh38
rs34990847 Chr.1: 83917700-83917700 on Build GRCh38
rs767721492 Chr.1: 83917562-83917562 on Build GRCh38
rs755398266 Chr.1: 83917590-83917590 on Build GRCh38
rs991941715 Chr.1: 83917694-83917694 on Build GRCh38
rs746049312 Chr.1: 83917728-83917741 on Build GRCh38
rs753051090 Chr.1: 83917595-83917595 on Build GRCh38
rs757457420 Chr.1: 83917604-83917604 on Build GRCh38
rs1016550974 Chr.1: 83917650-83917650 on Build GRCh38
rs373857845 Chr.1: 83917600-83917600 on Build GRCh38
rs1024799552 Chr.1: 83917695-83917695 on Build GRCh38
rs994363369 Chr.1: 83917742-83917742 on Build GRCh38
rs771851636 Chr.1: 83917626-83917626 on Build GRCh38
rs774686773 Chr.1: 83917726-83917726 on Build GRCh38
rs760298255 Chr.1: 83917640-83917640 on Build GRCh38
rs556159608 Chr.1: 83917705-83917705 on Build GRCh38
rs185633115 Chr.1: 83917617-83917617 on Build GRCh38
rs772839860 Chr.1: 83917569-83917569 on Build GRCh38
rs200358438 Chr.1: 83917627-83917627 on Build GRCh38
rs758871825 Chr.1: 83917727-83917727 on Build GRCh38
rs140584102 Chr.1: 83917680-83917680 on Build GRCh38
rs772246301 Chr.1: 83917729-83917729 on Build GRCh38
rs138340708 Chr.1: 83917619-83917619 on Build GRCh38
rs188748565 Chr.1: 83917722-83917722 on Build GRCh38
rs775858182 Chr.1: 83917736-83917737 on Build GRCh38
rs774211627 Chr.1: 83917622-83917622 on Build GRCh38
rs754417700 Chr.1: 83917587-83917587 on Build GRCh38
rs765944989 Chr.1: 83917669-83917669 on Build GRCh38
rs765775511 Chr.1: 83917583-83917583 on Build GRCh38
rs760246963 Chr.1: 83917579-83917579 on Build GRCh38

View Full Product Details