Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 41700176-41700403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INHBA
Entrez Gene ID
3624
Gene Name
inhibin beta A subunit
Gene Aliases
EDF, FRP
UniGene
Hs.28792
Gene Chromosome Location
Chr.7: 41685101-41710532 on Build GRCh38
Transcripts
NM_002192.2 XM_017012174.1 XM_017012175.1 XM_017012176.1 NR_027118.1 NR_027119.1

Gene Symbol
INHBA-AS1
Entrez Gene ID
285954
Gene Name
INHBA antisense RNA 1
Gene Aliases
-
UniGene
Hs.656869
Gene Chromosome Location
Chr.7: 41693916-41779378 on Build GRCh38
Transcripts
NM_002192.2 XM_017012174.1 XM_017012175.1 XM_017012176.1 NR_027118.1 NR_027119.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3832779 c.164T>G p.V55G Chr.7: 41700211-41700211 on Build GRCh38
6966561 c.66_67insC p.T23fs*84 Chr.7: 41700308-41700309 on Build GRCh38
48427 c.65C>A p.P22H Chr.7: 41700310-41700310 on Build GRCh38
6685874 c.91G>A p.A31T Chr.7: 41700284-41700284 on Build GRCh38
1450706 c.154C>T p.P52S Chr.7: 41700221-41700221 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147747702 Chr.7: 41700309-41700309 on Build GRCh38
rs755619740 Chr.7: 41700298-41700298 on Build GRCh38
rs972161023 Chr.7: 41700292-41700292 on Build GRCh38
rs745928293 Chr.7: 41700267-41700267 on Build GRCh38
rs762816927 Chr.7: 41700278-41700278 on Build GRCh38
rs777959895 Chr.7: 41700334-41700334 on Build GRCh38
rs771248041 Chr.7: 41700257-41700257 on Build GRCh38
rs779271157 Chr.7: 41700226-41700226 on Build GRCh38
rs534590694 Chr.7: 41700290-41700290 on Build GRCh38
rs145503621 Chr.7: 41700282-41700282 on Build GRCh38
rs755524019 Chr.7: 41700207-41700207 on Build GRCh38
rs757609397 Chr.7: 41700344-41700344 on Build GRCh38
rs373322295 Chr.7: 41700253-41700253 on Build GRCh38
rs984438766 Chr.7: 41700254-41700254 on Build GRCh38
rs749995930 Chr.7: 41700291-41700291 on Build GRCh38
rs758814874 Chr.7: 41700328-41700328 on Build GRCh38
rs201591674 Chr.7: 41700268-41700268 on Build GRCh38
rs376794102 Chr.7: 41700251-41700251 on Build GRCh38
rs149209111 Chr.7: 41700335-41700335 on Build GRCh38
rs933197708 Chr.7: 41700330-41700330 on Build GRCh38
rs775222611 Chr.7: 41700276-41700276 on Build GRCh38
rs765780968 Chr.7: 41700304-41700304 on Build GRCh38
rs761497837 Chr.7: 41700283-41700283 on Build GRCh38
rs953665038 Chr.7: 41700241-41700241 on Build GRCh38
rs919363715 Chr.7: 41700322-41700322 on Build GRCh38
rs1030538647 Chr.7: 41700213-41700213 on Build GRCh38
rs376838491 Chr.7: 41700281-41700281 on Build GRCh38
rs942081109 Chr.7: 41700285-41700285 on Build GRCh38
rs879251431 Chr.7: 41700312-41700312 on Build GRCh38
rs781558777 Chr.7: 41700381-41700381 on Build GRCh38
rs919035995 Chr.7: 41700201-41700201 on Build GRCh38
rs373520349 Chr.7: 41700259-41700259 on Build GRCh38
rs748615486 Chr.7: 41700243-41700243 on Build GRCh38

View Full Product Details