Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97930993-97931254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEMGN
Entrez Gene ID
55363
Gene Name
hemogen
Gene Aliases
CT155, EDAG, EDAG-1, NDR
UniGene
Hs.176626
Gene Chromosome Location
Chr.9: 97926791-97944915 on Build GRCh38
Transcripts
NM_018437.4 NM_197978.2 XM_005252086.1 XM_011518845.2 XM_011518846.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5912918 c.206G>A p.G69E Chr.9: 97931189-97931189 on Build GRCh38
7370971 c.394C>G p.H132D Chr.9: 97931001-97931001 on Build GRCh38
2793233 c.401C>T p.S134F Chr.9: 97930994-97930994 on Build GRCh38
2793235 c.322delA p.T108fs*7 Chr.9: 97931073-97931073 on Build GRCh38
6114621 c.335G>T p.G112V Chr.9: 97931060-97931060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs267602331 Chr.9: 97931040-97931040 on Build GRCh38
rs530693956 Chr.9: 97931132-97931132 on Build GRCh38
rs777381979 Chr.9: 97931024-97931024 on Build GRCh38
rs763084947 Chr.9: 97931206-97931206 on Build GRCh38
rs770629296 Chr.9: 97931039-97931039 on Build GRCh38
rs750485705 Chr.9: 97931209-97931209 on Build GRCh38
rs776710745 Chr.9: 97931198-97931198 on Build GRCh38
rs145386159 Chr.9: 97931088-97931088 on Build GRCh38
rs767265129 Chr.9: 97931178-97931178 on Build GRCh38
rs748934699 Chr.9: 97931033-97931033 on Build GRCh38
rs201982775 Chr.9: 97931159-97931159 on Build GRCh38
rs1030400684 Chr.9: 97931168-97931168 on Build GRCh38
rs766637508 Chr.9: 97931097-97931097 on Build GRCh38
rs759840748 Chr.9: 97931209-97931209 on Build GRCh38
rs990233097 Chr.9: 97931208-97931209 on Build GRCh38
rs142085732 Chr.9: 97931208-97931208 on Build GRCh38
rs370588490 Chr.9: 97931072-97931072 on Build GRCh38
rs745597622 Chr.9: 97931049-97931049 on Build GRCh38
rs1031519231 Chr.9: 97931185-97931186 on Build GRCh38
rs144511790 Chr.9: 97931084-97931084 on Build GRCh38
rs373987605 Chr.9: 97931130-97931130 on Build GRCh38
rs370929191 Chr.9: 97931094-97931094 on Build GRCh38
rs147951470 Chr.9: 97931145-97931145 on Build GRCh38
rs765066126 Chr.9: 97931073-97931073 on Build GRCh38
rs767972315 Chr.9: 97931133-97931133 on Build GRCh38
rs775245293 Chr.9: 97931068-97931068 on Build GRCh38
rs779943930 Chr.9: 97931177-97931177 on Build GRCh38
rs867323476 Chr.9: 97931189-97931189 on Build GRCh38
rs201990544 Chr.9: 97931105-97931107 on Build GRCh38
rs765203808 Chr.9: 97931074-97931074 on Build GRCh38
rs200834552 Chr.9: 97931047-97931047 on Build GRCh38
rs370935913 Chr.9: 97931135-97931135 on Build GRCh38
rs139664531 Chr.9: 97931063-97931063 on Build GRCh38
rs777135825 Chr.9: 97931146-97931146 on Build GRCh38
rs748709763 Chr.9: 97931017-97931017 on Build GRCh38
rs569650277 Chr.9: 97931110-97931110 on Build GRCh38
rs144965245 Chr.9: 97931105-97931105 on Build GRCh38
rs755555750 Chr.9: 97931023-97931023 on Build GRCh38
rs956296629 Chr.9: 97931201-97931201 on Build GRCh38
rs745544667 Chr.9: 97931171-97931171 on Build GRCh38
rs753287664 Chr.9: 97931149-97931149 on Build GRCh38
rs751768399 Chr.9: 97931072-97931073 on Build GRCh38
rs762091173 Chr.9: 97931093-97931093 on Build GRCh38
rs756709528 Chr.9: 97931140-97931140 on Build GRCh38
rs778499186 Chr.9: 97931163-97931163 on Build GRCh38
rs759349655 Chr.9: 97931091-97931091 on Build GRCh38
rs202156150 Chr.9: 97931173-97931173 on Build GRCh38
rs771154310 Chr.9: 97931207-97931207 on Build GRCh38
rs771744071 Chr.9: 97931172-97931172 on Build GRCh38
rs898151041 Chr.9: 97931092-97931092 on Build GRCh38
rs760717668 Chr.9: 97931070-97931070 on Build GRCh38

View Full Product Details