Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6452251-6452520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A23
Entrez Gene ID
79085
Gene Name
solute carrier family 25 member 23
Gene Aliases
APC2, MCSC2, SCaMC-3
UniGene
Hs.356231
Gene Chromosome Location
Chr.19: 6436081-6460221 on Build GRCh38
Transcripts
NM_024103.2 XM_011528274.1 XM_011528275.1 XM_011528276.1 XM_011528277.1 XM_011528278.1 XM_011528279.1 XM_011528280.1 XM_011528281.1 XM_011528282.1 XM_011528283.1 XM_011528284.1 XM_011528285.1 XM_017027285.1 XM_017027286.1 XM_017027287.1 XM_017027288.1 XM_017027289.1 XM_017027290.1 XM_017027291.1 XM_017027292.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4726811 c.1007G>A p.R336H Chr.19: 6452376-6452376 on Build GRCh38
3743183 c.963C>T p.C321C Chr.19: 6452420-6452420 on Build GRCh38
4726813 c.971G>A p.R324H Chr.19: 6452412-6452412 on Build GRCh38
6501471 c.951G>A p.G317G Chr.19: 6452432-6452432 on Build GRCh38
6780595 c.1046C>T p.A349V Chr.19: 6452337-6452337 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61729423 Chr.19: 6452412-6452412 on Build GRCh38
rs982637646 Chr.19: 6452305-6452305 on Build GRCh38
rs952458735 Chr.19: 6452494-6452494 on Build GRCh38
rs773613499 Chr.19: 6452327-6452327 on Build GRCh38
rs746153495 Chr.19: 6452501-6452501 on Build GRCh38
rs771153945 Chr.19: 6452338-6452338 on Build GRCh38
rs141724728 Chr.19: 6452336-6452336 on Build GRCh38
rs140701489 Chr.19: 6452420-6452420 on Build GRCh38
rs374320646 Chr.19: 6452296-6452296 on Build GRCh38
rs772084423 Chr.19: 6452481-6452482 on Build GRCh38
rs147601819 Chr.19: 6452375-6452375 on Build GRCh38
rs772901772 Chr.19: 6452465-6452465 on Build GRCh38
rs377043212 Chr.19: 6452466-6452466 on Build GRCh38
rs751635874 Chr.19: 6452284-6452284 on Build GRCh38
rs182419131 Chr.19: 6452314-6452314 on Build GRCh38
rs756646377 Chr.19: 6452474-6452474 on Build GRCh38
rs173229 Chr.19: 6452294-6452294 on Build GRCh38
rs149780316 Chr.19: 6452468-6452468 on Build GRCh38
rs1000790478 Chr.19: 6452326-6452326 on Build GRCh38
rs61729395 Chr.19: 6452315-6452315 on Build GRCh38
rs267605748 Chr.19: 6452390-6452390 on Build GRCh38
rs764141860 Chr.19: 6452360-6452360 on Build GRCh38
rs745948644 Chr.19: 6452447-6452447 on Build GRCh38
rs766522267 Chr.19: 6452493-6452493 on Build GRCh38
rs748515868 Chr.19: 6452320-6452320 on Build GRCh38
rs776646174 Chr.19: 6452369-6452369 on Build GRCh38
rs145400081 Chr.19: 6452405-6452405 on Build GRCh38
rs185375372 Chr.19: 6452402-6452402 on Build GRCh38
rs369720632 Chr.19: 6452374-6452374 on Build GRCh38
rs781618762 Chr.19: 6452318-6452318 on Build GRCh38
rs932350093 Chr.19: 6452451-6452451 on Build GRCh38
rs140513558 Chr.19: 6452388-6452388 on Build GRCh38
rs757577296 Chr.19: 6452491-6452491 on Build GRCh38
rs750114314 Chr.19: 6452299-6452299 on Build GRCh38
rs749629974 Chr.19: 6452485-6452485 on Build GRCh38
rs758014446 Chr.19: 6452377-6452377 on Build GRCh38
rs769758582 Chr.19: 6452359-6452359 on Build GRCh38
rs760741163 Chr.19: 6452489-6452491 on Build GRCh38
rs779482157 Chr.19: 6452312-6452312 on Build GRCh38
rs374257352 Chr.19: 6452419-6452419 on Build GRCh38
rs779144308 Chr.19: 6452499-6452499 on Build GRCh38
rs761840721 Chr.19: 6452372-6452372 on Build GRCh38
rs561972759 Chr.19: 6452298-6452298 on Build GRCh38
rs752171507 Chr.19: 6452472-6452472 on Build GRCh38
rs199968517 Chr.19: 6452413-6452413 on Build GRCh38
rs778294050 Chr.19: 6452479-6452479 on Build GRCh38
rs967989404 Chr.19: 6452323-6452323 on Build GRCh38
rs148491192 Chr.19: 6452321-6452321 on Build GRCh38
rs267605746 Chr.19: 6452344-6452344 on Build GRCh38
rs761892055 Chr.19: 6452292-6452292 on Build GRCh38
rs775700306 Chr.19: 6452454-6452454 on Build GRCh38
rs773455273 Chr.19: 6452486-6452487 on Build GRCh38
rs753709495 Chr.19: 6452297-6452297 on Build GRCh38
rs975142731 Chr.19: 6452282-6452282 on Build GRCh38
rs774326452 Chr.19: 6452345-6452345 on Build GRCh38
rs754609735 Chr.19: 6452469-6452469 on Build GRCh38
rs4807869 Chr.19: 6452455-6452455 on Build GRCh38
rs267605747 Chr.19: 6452389-6452389 on Build GRCh38
rs1000951036 Chr.19: 6452408-6452408 on Build GRCh38
rs1008467654 Chr.19: 6452448-6452448 on Build GRCh38
rs754143737 Chr.19: 6452494-6452495 on Build GRCh38
rs377691247 Chr.19: 6452452-6452452 on Build GRCh38
rs758059613 Chr.19: 6452311-6452311 on Build GRCh38
rs767063113 Chr.19: 6452470-6452470 on Build GRCh38
rs774576513 Chr.19: 6452432-6452432 on Build GRCh38
rs765193901 Chr.19: 6452462-6452462 on Build GRCh38
rs778038007 Chr.19: 6452414-6452414 on Build GRCh38
rs765948848 Chr.19: 6452467-6452467 on Build GRCh38

View Full Product Details