Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5210574-5210799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRS
Entrez Gene ID
5802
Gene Name
protein tyrosine phosphatase, receptor type S
Gene Aliases
PTPSIGMA, R-PTP-S, R-PTP-sigma
UniGene
Hs.744928
Gene Chromosome Location
Chr.19: 5205503-5340803 on Build GRCh38
Transcripts
NM_002850.3 NM_130853.2 NM_130854.2 NM_130855.2 XM_005259600.2 XM_005259606.2 XM_005259607.2 XM_005259609.1 XM_005259610.1 XM_011528157.2 XM_011528158.2 XM_017027065.1 XM_017027066.1 XM_017027067.1 XM_017027068.1 XM_017027069.1 XM_017027070.1 XM_017027071.1 XM_017027072.1 XM_017027073.1 XM_017027074.1 XM_017027075.1 XM_017027076.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4979956 c.5256G>A p.A1752A Chr.19: 5210784-5210784 on Build GRCh38
6930235 c.5273C>T p.A1758V Chr.19: 5210767-5210767 on Build GRCh38
3773129 c.5326G>A p.V1776M Chr.19: 5210714-5210714 on Build GRCh38
2926131 c.5301G>T p.M1767I Chr.19: 5210739-5210739 on Build GRCh38
148463 c.5269C>T p.L1757L Chr.19: 5210771-5210771 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776795641 Chr.19: 5210683-5210683 on Build GRCh38
rs762745270 Chr.19: 5210773-5210773 on Build GRCh38
rs754495645 Chr.19: 5210614-5210614 on Build GRCh38
rs1143698 Chr.19: 5210771-5210771 on Build GRCh38
rs372417196 Chr.19: 5210618-5210618 on Build GRCh38
rs374038876 Chr.19: 5210718-5210718 on Build GRCh38
rs547394205 Chr.19: 5210772-5210772 on Build GRCh38
rs760692982 Chr.19: 5210669-5210669 on Build GRCh38
rs2302224 Chr.19: 5210611-5210611 on Build GRCh38
rs576139901 Chr.19: 5210617-5210617 on Build GRCh38
rs370307369 Chr.19: 5210649-5210649 on Build GRCh38
rs746423364 Chr.19: 5210738-5210738 on Build GRCh38
rs1022831616 Chr.19: 5210634-5210634 on Build GRCh38
rs201047108 Chr.19: 5210757-5210757 on Build GRCh38
rs776812271 Chr.19: 5210662-5210662 on Build GRCh38
rs759512233 Chr.19: 5210703-5210703 on Build GRCh38
rs777508314 Chr.19: 5210621-5210621 on Build GRCh38
rs747820983 Chr.19: 5210616-5210616 on Build GRCh38
rs374377699 Chr.19: 5210767-5210767 on Build GRCh38
rs770699501 Chr.19: 5210758-5210758 on Build GRCh38
rs768674983 Chr.19: 5210743-5210743 on Build GRCh38
rs758125715 Chr.19: 5210607-5210607 on Build GRCh38
rs762760744 Chr.19: 5210692-5210692 on Build GRCh38
rs199914555 Chr.19: 5210667-5210667 on Build GRCh38
rs779307747 Chr.19: 5210650-5210650 on Build GRCh38
rs774182734 Chr.19: 5210759-5210759 on Build GRCh38
rs112380103 Chr.19: 5210596-5210596 on Build GRCh38
rs377459393 Chr.19: 5210610-5210610 on Build GRCh38
rs116345982 Chr.19: 5210714-5210714 on Build GRCh38
rs779302415 Chr.19: 5210722-5210722 on Build GRCh38
rs746359177 Chr.19: 5210657-5210657 on Build GRCh38
rs1143699 Chr.19: 5210751-5210751 on Build GRCh38
rs780361257 Chr.19: 5210739-5210739 on Build GRCh38
rs756926448 Chr.19: 5210778-5210778 on Build GRCh38
rs750235235 Chr.19: 5210604-5210604 on Build GRCh38
rs747125571 Chr.19: 5210741-5210741 on Build GRCh38
rs759194375 Chr.19: 5210763-5210763 on Build GRCh38
rs1031559702 Chr.19: 5210686-5210686 on Build GRCh38
rs752263180 Chr.19: 5210715-5210715 on Build GRCh38
rs766174955 Chr.19: 5210693-5210693 on Build GRCh38
rs780723497 Chr.19: 5210615-5210615 on Build GRCh38
rs767493286 Chr.19: 5210765-5210765 on Build GRCh38
rs72981058 Chr.19: 5210624-5210624 on Build GRCh38
rs978677881 Chr.19: 5210651-5210651 on Build GRCh38
rs547580599 Chr.19: 5210644-5210644 on Build GRCh38
rs753489907 Chr.19: 5210719-5210719 on Build GRCh38
rs371091292 Chr.19: 5210766-5210766 on Build GRCh38
rs768873099 Chr.19: 5210676-5210676 on Build GRCh38
rs751492887 Chr.19: 5210699-5210699 on Build GRCh38
rs1049873225 Chr.19: 5210779-5210779 on Build GRCh38

View Full Product Details