Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151816581-151816854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RORC
Entrez Gene ID
6097
Gene Name
RAR related orphan receptor C
Gene Aliases
IMD42, NR1F3, RORG, RZR-GAMMA, RZRG, TOR
UniGene
Hs.256022
Gene Chromosome Location
Chr.1: 151806071-151832450 on Build GRCh38
Transcripts
NM_001001523.1 NM_005060.3 XM_006711484.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1659885 c.166C>T p.R56C Chr.1: 151816796-151816796 on Build GRCh38
3399700 c.425T>C p.L142P Chr.1: 151816699-151816699 on Build GRCh38
5377621 c.298+1G>A p.? Chr.1: 151816663-151816663 on Build GRCh38
5377623 c.165C>T p.F55F Chr.1: 151816797-151816797 on Build GRCh38
5538336 c.239G>A p.R80Q Chr.1: 151816723-151816723 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201345613 Chr.1: 151816815-151816815 on Build GRCh38
rs777497402 Chr.1: 151816757-151816757 on Build GRCh38
rs374584720 Chr.1: 151816770-151816770 on Build GRCh38
rs34830957 Chr.1: 151816776-151816776 on Build GRCh38
rs199891898 Chr.1: 151816764-151816764 on Build GRCh38
rs1050630219 Chr.1: 151816716-151816716 on Build GRCh38
rs773795049 Chr.1: 151816627-151816627 on Build GRCh38
rs201334909 Chr.1: 151816683-151816683 on Build GRCh38
rs777188216 Chr.1: 151816707-151816707 on Build GRCh38
rs940439345 Chr.1: 151816604-151816604 on Build GRCh38
rs745389017 Chr.1: 151816647-151816647 on Build GRCh38
rs751829653 Chr.1: 151816736-151816736 on Build GRCh38
rs765622050 Chr.1: 151816820-151816820 on Build GRCh38
rs763000024 Chr.1: 151816835-151816835 on Build GRCh38
rs772506661 Chr.1: 151816647-151816648 on Build GRCh38
rs531998565 Chr.1: 151816607-151816607 on Build GRCh38
rs750400604 Chr.1: 151816730-151816730 on Build GRCh38
rs200944681 Chr.1: 151816717-151816717 on Build GRCh38
rs201956211 Chr.1: 151816834-151816834 on Build GRCh38
rs200519552 Chr.1: 151816653-151816653 on Build GRCh38
rs901597268 Chr.1: 151816698-151816698 on Build GRCh38
rs546157871 Chr.1: 151816684-151816684 on Build GRCh38
rs201820724 Chr.1: 151816645-151816645 on Build GRCh38
rs776802205 Chr.1: 151816795-151816795 on Build GRCh38
rs201003278 Chr.1: 151816629-151816629 on Build GRCh38
rs745951980 Chr.1: 151816623-151816624 on Build GRCh38
rs746722517 Chr.1: 151816783-151816783 on Build GRCh38
rs758504357 Chr.1: 151816732-151816732 on Build GRCh38
rs778919253 Chr.1: 151816773-151816773 on Build GRCh38
rs200047866 Chr.1: 151816651-151816651 on Build GRCh38
rs143596741 Chr.1: 151816774-151816774 on Build GRCh38
rs913307383 Chr.1: 151816782-151816782 on Build GRCh38
rs368393221 Chr.1: 151816722-151816722 on Build GRCh38
rs142141845 Chr.1: 151816709-151816709 on Build GRCh38
rs773730516 Chr.1: 151816821-151816821 on Build GRCh38
rs755871920 Chr.1: 151816756-151816756 on Build GRCh38
rs201373806 Chr.1: 151816775-151816775 on Build GRCh38
rs202196367 Chr.1: 151816746-151816746 on Build GRCh38
rs993873318 Chr.1: 151816616-151816616 on Build GRCh38
rs200449406 Chr.1: 151816785-151816785 on Build GRCh38
rs749237284 Chr.1: 151816761-151816761 on Build GRCh38

View Full Product Details